Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03664588_s1
          See other LOC100128591 GE Assays ›
          Gene Symbol:
          LOC100128591
          Gene Name:
          uncharacterized LOC100128591
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03664588_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100128591
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK128705.1 - 1 3450 89
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DB352726.1 - 1 Chr. 20 - 46331942 - 46332512 [+] on Build GRCh38 89

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cadherin 22 CDH22 C20orf25,
          dJ998H6.1
          -23.1kb Sense Chr. 20 - 46173733 - 46308498 on NCBI Build GRCh38
          Downstream solute carrier family 35 member C2 SLC35C2 BA394O2.1,
          C20orf5,
          CGI-15,
          OVCOV1
          13.7kb Sense Chr. 20 - 46349528 - 46364461 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31838828 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190877 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190925 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190898 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190894 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31838898 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31838842 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31838844 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190920 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190865 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17195523 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190875 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190924 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190851 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190860 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190911 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190855 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190917 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31838854 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190881 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190847 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190907 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17195520 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190914 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190871 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17195534 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190876 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190854 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31838850 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190901 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31838833 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31838841 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190846 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190908 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190888 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31838893 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190863 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190905 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190897 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31838824 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17195517 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31838878 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190844 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190923 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31838887 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31838859 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190893 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190853 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190845 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31838880 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31838847 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31838846 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17195518 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190843 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31838867 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31838861 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31838818 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190915 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31838858 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190896 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190885 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190850 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31838890 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190922 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190858 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17195519 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190904 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190927 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31838837 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190873 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190891 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190895 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190903 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190884 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190906 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31838869 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11190921 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 815966 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 131140 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 294810 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 292680 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 413980 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 397332 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 204365 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 209918 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3402943 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 2247 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3402943 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 172232 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P34723939 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03525117 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03525117 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0500007418.hg.1 ENST00000506435.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007418.hg.1 ENST00000506435.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007418.hg.1 ENST00000506435.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007418.hg.1 ENST00000506435.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007418.hg.1 ENST00000506435.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007418.hg.1 ENST00000506435.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007418.hg.1 ENST00000506435.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007418.hg.1 ENST00000506435.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007418.hg.1 ENST00000506435.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007418.hg.1 ENST00000506435.1 Ensembl
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3190442 - -
          HumanHT-12 V4.0 expression beadchip ILMN_3190442 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0