Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03665280_m1
          See other LOC387810 GE Assays ›
          Gene Symbol:
          LOC387810
          Gene Name:
          uncharacterized LOC387810
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 112534220 - 112555802 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03665280_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC387810
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_104155.1 3 - 4 436 98
          GenBank mRNA BC070394.1 - 3 - 4 436 98
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BU506733.1 - 3 - 4 438 Chr. 11 - 112535559 - 112555802 [-] on Build GRCh38 98

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC101928823 LOC101928823 Anti-sense Chr. 11 - 112482232 - 112541916 on NCBI Build GRCh38
          Downstream uncharacterized LOC101928847 LOC101928847 410.9kb Sense Chr. 11 - 112959279 - 112963460 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100012332.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033960 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460749 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033986 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16502550 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16502529 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460736 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033987 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034041 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033971 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460665 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033941 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460654 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460675 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033984 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033996 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033950 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034081 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460739 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460753 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460688 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034024 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460772 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033982 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033977 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034005 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034006 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460761 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033994 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460747 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460670 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460705 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460732 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16502530 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16502548 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460720 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034013 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460744 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460730 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460782 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460776 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034012 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16502553 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033953 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034044 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034063 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460656 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034053 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034017 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460774 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460653 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460710 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460791 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033983 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033961 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460669 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033956 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16502535 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033942 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460778 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034060 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034002 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460691 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460775 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16502542 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033978 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460727 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034007 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034018 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034037 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034028 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034036 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033992 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460676 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460777 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460760 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034050 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034000 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460719 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034030 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460664 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033980 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460741 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034004 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034001 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460671 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033938 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034073 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034061 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460682 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460793 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460709 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460773 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16502545 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460734 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033991 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034011 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460790 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460659 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034022 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460763 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460762 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034026 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033939 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033970 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460643 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16502541 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460696 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460698 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033948 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460764 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034072 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460765 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16502552 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460700 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460667 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034031 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034052 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034074 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460646 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460785 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034080 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034047 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460672 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033976 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034014 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08033949 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460766 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460733 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034020 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460689 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08034077 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28460716 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 37092 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 492520 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 813465 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 51829 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 20912 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 657806 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 972470 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3589420 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 52172 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P19373493 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P19373493 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3589420 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P19373493 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 58636 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19373493 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19373502 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P11242041 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P11245215 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P19373493 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1100012332.hg.1 ENST00000525984.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012332.hg.1 ENST00000525984.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012332.hg.1 ENST00000525984.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012332.hg.1 ENST00000525984.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012332.hg.1 ENST00000525984.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012332.hg.1 ENST00000525984.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012332.hg.1 ENST00000525984.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012332.hg.1 ENST00000525984.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012332.hg.1 NR_104155.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012332.hg.1 NR_104155.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012332.hg.1 NR_104155.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012332.hg.1 NR_104155.1 Refseq
          Affymetrix Human Clariom D Assay TC1100012332.hg.1 ENST00000525984.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012332.hg.1 ENST00000525984.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012332.hg.1 ENST00000525984.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012332.hg.1 ENST00000525984.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012332.hg.1 ENST00000525984.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012332.hg.1 ENST00000525984.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012332.hg.1 ENST00000525984.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012332.hg.1 ENST00000525984.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012332.hg.1 NR_104155.1 Refseq
          Affymetrix Human Clariom D Assay TC1100012332.hg.1 NR_104155.1 Refseq
          Affymetrix Human Clariom D Assay TC1100012332.hg.1 NR_104155.1 Refseq
          Affymetrix Human Clariom D Assay TC1100012332.hg.1 NR_104155.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100194732.hg.1 ENST00000525984.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100194732.hg.1 ENST00000525984.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100194732.hg.1 NR_104155.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100194732.hg.1 TC1100012332.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100194732.hg.1 NR_104155.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100194732.hg.1 ENST00000525984.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100194732.hg.1 ENST00000525984.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0