Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03665345_s1
          See other LOC729683 GE Assays ›
          Gene Symbol:
          LOC729683
          Gene Name:
          uncharacterized LOC729683
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 63700338 - 63702685 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03665345_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC729683
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046273.1 1 1856 68
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BG341177.1 - 1 462 Chr. 17 - 63701732 - 63702380 [+] on Build GRCh38 68
          GenBankmRNA BX342850.2 - 1 Chr. 17 - 63701650 - 63702666 [-] on Build GRCh38 68
          GenBankmRNA BX107071.1 - 1 46 Chr. 17 - 63702149 - 63702614 [+] on Build GRCh38 68

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream LIM domain containing 2 LIMD2 Sense Chr. 17 - 63695889 - 63701222 on NCBI Build GRCh38
          Downstream STE20-related kinase adaptor alpha STRADA LYK5,
          NY-BR-96,
          PMSE,
          STRAD,
          Stlk
          166.0 Sense Chr. 17 - 63702832 - 63741970 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700008569.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P145010 - -
          SurePrint G3 Human GE Microarray G4851A A_32_P145010 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P145010 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P49631 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 236497_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 236614_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1700008569.hg.1 ENST00000623228.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008569.hg.1 NR_046273.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008569.hg.1 NR_046273.1 Refseq
          Affymetrix Human Clariom D Assay TC1700008569.hg.1 ENST00000623228.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008569.hg.1 NR_046273.1 Refseq
          Affymetrix Human Clariom D Assay TC1700008569.hg.1 NR_046273.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700164063.hg.1 NR_046273.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700164063.hg.1 TC1700008569.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700164063.hg.1 NR_046273.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700164064.hg.1 ENST00000623228.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700164064.hg.1 NR_046273.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700164064.hg.1 TC1700008569.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700164064.hg.1 ENST00000623228.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700164064.hg.1 NR_046273.1 Refseq
          Human Genome U133 Plus 2.0 Array 236614_PM_at - -
          Human Genome U133 Plus 2.0 Array 236497_at - -
          Human Genome U133 Plus 2.0 Array 236497_PM_at - -
          Human Genome U133 Plus 2.0 Array 236614_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236497_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236614_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236614_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236497_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236614_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236497_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236497_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236614_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236497_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236497_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236614_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236497_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236614_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236614_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236497_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236614_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236497_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236497_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236497_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236614_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236614_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236614_at5 - -
          Human Genome U95 Set 81465_PM_at - -
          Human Genome U95 Set 81465_at - -
          Human Genome U95 Set 83614_at - -
          Human Genome U95 Set 83614_PM_at - -
          Human Genome U133 Set 236614_PM_at - -
          Human Genome U133 Set 236497_at - -
          Human Genome U133 Set 236614_at - -
          Human Genome U133 Set 236497_PM_at - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3298163 CR594811.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3298163 CR594811.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0