Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT HG-U133+ PM Array Plate |
1553374_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011391.hg.1 |
ENST00000319051.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011391.hg.1 |
ENST00000319051.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011391.hg.1 |
ENST00000319051.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011391.hg.1 |
ENST00000622091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011391.hg.1 |
ENST00000622091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011391.hg.1 |
ENST00000622091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011391.hg.1 |
ENST00000319051.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011391.hg.1 |
ENST00000319051.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011391.hg.1 |
ENST00000319051.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011391.hg.1 |
ENST00000622091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011391.hg.1 |
ENST00000622091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011391.hg.1 |
ENST00000622091.1 |
Ensembl |
Human Genome U133 Plus 2.0 Array |
1553374_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1553374_PM_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1553374_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1553374_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1553374_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1553374_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1553374_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1553374_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1553374_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1553374_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1553374_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1553374_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1553374_at6 |
- |
- |