Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03667035_s1
          See other LOC100131508 GE Assays ›
          Gene Symbol:
          LOC100131508
          Gene Name:
          PRO2122
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03667035_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100131508
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF130064.2 - 1 763 80

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream branched chain keto acid dehydrogenase E1 subunit beta BCKDHB BCKDE1B,
          BCKDH E1-beta,
          E1B
          -1.2mb Anti-sense Chr. 6 - 80106610 - 80469088 on NCBI Build GRCh38
          Downstream family with sequence similarity 46 member A FAM46A C6orf37,
          XTP11
          191.6kb Sense Chr. 6 - 81745730 - 81752711 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AF130064.2 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051307 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051241 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051262 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051298 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051242 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051300 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04851500 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051296 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051264 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15834680 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051313 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04851516 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051251 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051236 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051230 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051286 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051281 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051252 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04851482 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04851463 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04851498 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051239 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051277 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04851442 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051248 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051255 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04851491 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051302 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051250 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04851528 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051226 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051306 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04851474 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051279 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051245 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051316 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051288 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051244 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04851450 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051238 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051219 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15834687 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051233 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051310 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051287 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051293 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04851452 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051229 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051247 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051271 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04851444 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04851502 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051274 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051223 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04851527 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04851443 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04851520 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051231 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051284 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04851530 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04851507 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051305 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051249 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051285 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051240 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051270 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051291 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04851441 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04851524 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051258 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051280 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04851488 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051292 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04851525 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051311 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04851519 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051309 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051314 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25051263 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 51022 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P239718 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 43220 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P239718 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P239718 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array XM_001725343_at - -
          Affymetrix Human Gene 1.0 ST Array XM_001720073_at - -
          Affymetrix Human Gene 1.0 ST Array XM_001719407_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0600008648.hg.1 ENST00000614959.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600008648.hg.1 ENST00000614959.1 Ensembl
          Human Genome U133 Plus 2.0 Array 224351_at - -
          Human Genome U133 Plus 2.0 Array 224351_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224351_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224351_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224351_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224351_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224351_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224351_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224351_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224351_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224351_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224351_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224351_at11 - -
          Human Genome U133 Set 224351_PM_at - -
          Human Genome U133 Set 224351_at - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1843399 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 540368 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_110471 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1843399 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-8696d955b-mh7vc:80/100.66.135.11:80.
          git-commit: b6496d4449ebd8b46e03b29da6eff4841d0bfc13
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline