Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03667384_s1
          See other LOC90784 GE Assays ›
          Gene Symbol:
          LOC90784
          Gene Name:
          uncharacterized LOC90784
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 86020216 - 86023868 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03667384_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC90784
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026984.1 1 2401 67
          GenBank mRNA AK001612.1 - 1 2045 67
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BQ014804.1 - 1 Chr. 2 - 86020933 - 86021560 [+] on Build GRCh38 67
          GenBankmRNA BU859578.1 - 1 155 Chr. 2 - 86020768 - 86021622 [-] on Build GRCh38 67

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ST3GAL5 antisense RNA 1 (head to head) ST3GAL5-AS1 -128.6kb Anti-sense Chr. 2 - 85889280 - 85892252 on NCBI Build GRCh38
          Downstream RNA polymerase I subunit A POLR1A A190,
          AFDCIN,
          RPA1,
          RPA194,
          RPO1-4,
          RPO14
          1.7kb Sense Chr. 2 - 86026328 - 86106155 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200016701.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02002050.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01332612 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21415294 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01332634 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01332617 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21415296 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21415316 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01332639 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01332633 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21415293 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01332620 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21415289 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01332630 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21415288 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21415305 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21415287 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21415291 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01332638 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01332629 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21415300 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01332623 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01332622 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01332627 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 270279 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 264588 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3271480 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4905 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4904 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 4150 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3271480 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3271480 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01332660 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21415360 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21415356 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15263979 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21415361 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01332666 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01332681 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01332674 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01332669 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01332664 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01332670 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01332661 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01332684 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01332679 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01332682 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01332679 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01332684 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01332666 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01332682 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01332661 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21415356 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01332681 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21415360 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01332664 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01332660 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01332670 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01332674 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01332669 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15263979 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21415361 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00421882 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00421883 - -
          Agilent Human 2 cDNA Microarray (G4101A) 1274773 - -
          Agilent Human 2 cDNA Microarray (G4101A) 1274773 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332645 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332612 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332624 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332637 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332639 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15263966 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332633 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332647 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332678 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332656 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332664 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332666 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332617 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15263974 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332629 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15263976 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332648 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332640 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332620 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332669 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332650 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332627 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332631 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332614 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332671 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15263979 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332622 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332661 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332644 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332674 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332652 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332655 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332676 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332613 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15263977 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15263975 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332635 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15263971 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01332681 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00421882 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 228515_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR02032812.hg.1 NR_026984.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02032812.hg.1 TC02002050.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200227061.hg.1 NR_026984.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200227061.hg.1 TC0200016701.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200227061.hg.1 NR_026984.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200227063.hg.1 NR_026984.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200227063.hg.1 TC0200016701.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200227063.hg.1 NR_026984.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200227062.hg.1 NR_026984.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200227062.hg.1 TC0200016701.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200227062.hg.1 NR_026984.1 Refseq
          Human Genome U133 Plus 2.0 Array 228515_PM_at - -
          Human Genome U133 Plus 2.0 Array 228515_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228515_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228515_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228515_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228515_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228515_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228515_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228515_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228515_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228515_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228515_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228515_at7 - -
          Human Genome U95 Set 45463_PM_at - -
          Human Genome U95 Set 45463_at - -
          Human Genome U95 Set 61086_PM_at - -
          Human Genome U95 Set 61086_at - -
          Human Genome U95 Set 51233_PM_at - -
          Human Genome U95 Set 51233_at - -
          Human Genome U133 Set 228515_at - -
          Human Genome U133 Set 228515_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A013958 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A013958 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 233948 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0