Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03668393_s1
          See other LOC102724002 GE Assays ›
          Gene Symbol:
          LOC102724002
          Gene Name:
          uncharacterized LOC102724002
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03668393_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC102724002
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK025166.1 - 1 1749 89
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BF110595.1 - 1 Chr. 10 - 96995500 - 96995956 [-] on Build GRCh38 89
          GenBankmRNA AI089663.1 - 1 Chr. 10 - 96995480 - 96995955 [-] on Build GRCh38 89
          GenBankmRNA AA913401.1 - 1 Chr. 10 - 96995594 - 96995956 [-] on Build GRCh38 89
          GenBankmRNA AI382690.1 - 1 Chr. 10 - 96995674 - 96995956 [-] on Build GRCh38 89
          GenBankmRNA BM664328.1 - 1 Chr. 10 - 96995330 - 96995956 [-] on Build GRCh38 89
          GenBankmRNA DB549826.1 - 1 261 Chr. 10 - 96995479 - 96995954 [+] on Build GRCh38 89
          GenBankmRNA BF108543.1 - 1 Chr. 10 - 96995518 - 96995956 [-] on Build GRCh38 89
          GenBankmRNA AW614954.1 - 1 Chr. 10 - 96995591 - 96995953 [-] on Build GRCh38 89
          GenBankmRNA AW295840.1 - 1 Chr. 10 - 96995482 - 96995956 [-] on Build GRCh38 89
          GenBankmRNA AI167345.1 - 1 Chr. 10 - 96995523 - 96995956 [-] on Build GRCh38 89
          GenBankmRNA AI375611.1 - 1 Chr. 10 - 96995482 - 96995954 [-] on Build GRCh38 89
          GenBankmRNA BF108501.1 - 1 Chr. 10 - 96995511 - 96995956 [-] on Build GRCh38 89
          GenBankmRNA BI495649.1 - 1 261 Chr. 10 - 96995484 - 96995956 [+] on Build GRCh38 89
          GenBankmRNA AI375613.1 - 1 Chr. 10 - 96995597 - 96995954 [-] on Build GRCh38 89
          GenBankmRNA BF941449.1 - 1 Chr. 10 - 96995511 - 96995954 [-] on Build GRCh38 89
          GenBankmRNA AA258792.1 - 1 Chr. 10 - 96995566 - 96995954 [-] on Build GRCh38 89
          GenBankmRNA BX496982.1 - 1 373 Chr. 10 - 96995366 - 96995951 [+] on Build GRCh38 89
          GenBankmRNA BE549670.1 - 1 Chr. 10 - 96995436 - 96995953 [-] on Build GRCh38 89
          GenBankmRNA BF444964.1 - 1 Chr. 10 - 96995503 - 96995956 [-] on Build GRCh38 89
          GenBankmRNA AA947690.1 - 1 Chr. 10 - 96995565 - 96995953 [-] on Build GRCh38 89
          GenBankmRNA AW207892.1 - 1 Chr. 10 - 96995359 - 96995956 [-] on Build GRCh38 89
          GenBankmRNA AA533444.1 - 1 Chr. 10 - 96995536 - 96995925 [-] on Build GRCh38 89
          GenBankmRNA CA310504.1 - 1 Chr. 10 - 96995211 - 96995956 [-] on Build GRCh38 89
          GenBankmRNA AI681306.1 - 1 Chr. 10 - 96995402 - 96995953 [-] on Build GRCh38 89
          GenBankmRNA BI495648.1 - 1 Chr. 10 - 96995484 - 96995956 [-] on Build GRCh38 89
          GenBankmRNA BM681184.1 - 1 Chr. 10 - 96995284 - 96995956 [-] on Build GRCh38 89
          GenBankmRNA BE221986.1 - 1 Chr. 10 - 96995482 - 96995954 [-] on Build GRCh38 89
          GenBankmRNA AI280200.1 - 1 Chr. 10 - 96995489 - 96995954 [-] on Build GRCh38 89
          GenBankmRNA BU742285.1 - 1 Chr. 10 - 96995279 - 96995956 [-] on Build GRCh38 89
          GenBankmRNA BE858631.1 - 1 Chr. 10 - 96995400 - 96995954 [-] on Build GRCh38 89
          GenBankmRNA AI702215.1 - 1 Chr. 10 - 96995328 - 96995953 [-] on Build GRCh38 89
          GenBankmRNA BM990121.1 - 1 Chr. 10 - 96995312 - 96995956 [-] on Build GRCh38 89
          GenBankmRNA AI097242.1 - 1 Chr. 10 - 96995573 - 96995955 [-] on Build GRCh38 89

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream chromosome 10 open reading frame 12 C10orf12 -8.2kb Sense Chr. 10 - 96981284 - 96985828 on NCBI Build GRCh38
          Downstream slit guidance ligand 1 SLIT1 MEGF4,
          SLIL1,
          SLIT-1,
          SLIT3
          2.1kb Anti-sense Chr. 10 - 96998038 - 97185926 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1000008559.hg.1 ENST00000421806.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008559.hg.1 ENST00000421806.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008559.hg.1 ENST00000421806.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008559.hg.1 ENST00000421806.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0