Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004343.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004343.hg.1 |
TC18000428.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004345.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004345.hg.1 |
TC18000428.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008343.hg.1 |
ENST00000391065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008343.hg.1 |
ENST00000391065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008343.hg.1 |
ENST00000391065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008343.hg.1 |
ENST00000391065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008343.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008343.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008343.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008343.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008343.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008343.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008343.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008343.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008343.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008343.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008343.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008343.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008343.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008343.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008343.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008343.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004346.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004346.hg.1 |
TC18000428.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004344.hg.1 |
ENST00000391065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004344.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004344.hg.1 |
TC18000428.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006950.hg.1 |
ENST00000583852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006950.hg.1 |
ENST00000583852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156892.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156892.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156892.hg.1 |
TC1800008343.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800156892.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156892.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156896.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156896.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156896.hg.1 |
TC1800008343.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800156896.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156896.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156898.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156898.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156898.hg.1 |
TC1800008343.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800156898.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156898.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156891.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156891.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156891.hg.1 |
TC1800008343.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800156891.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156891.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156899.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156899.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156899.hg.1 |
TC1800008343.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800156899.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156899.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006950.hg.1 |
ENST00000583852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006950.hg.1 |
ENST00000583852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156897.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156897.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156897.hg.1 |
TC1800008343.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800156897.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156897.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008343.hg.1 |
ENST00000391065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008343.hg.1 |
ENST00000391065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008343.hg.1 |
ENST00000391065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008343.hg.1 |
ENST00000391065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008343.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008343.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008343.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008343.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008343.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008343.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008343.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008343.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008343.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008343.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008343.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008343.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008343.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008343.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008343.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008343.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156894.hg.1 |
ENST00000391065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156894.hg.1 |
ENST00000391065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156894.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156894.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156894.hg.1 |
TC1800008343.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800156894.hg.1 |
ENST00000391065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156894.hg.1 |
ENST00000391065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156894.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156894.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156895.hg.1 |
ENST00000391065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156895.hg.1 |
ENST00000391065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156895.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156895.hg.1 |
NR_024259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156895.hg.1 |
TC1800008343.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800156895.hg.1 |
ENST00000391065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156895.hg.1 |
ENST00000391065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156895.hg.1 |
ENST00000579458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156895.hg.1 |
NR_024259.1 |
Refseq |