Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03673327_s1
          See other UCKL1-AS1 GE Assays ›
          Gene Symbol:
          UCKL1-AS1
          Gene Name:
          UCKL1 antisense RNA 1
          Gene Aliases:
          UCKL1-AS, UCKL1AS, UCKL1OS
          Chromosome Location:
          Chr.20: 63953384 - 63956985 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03673327_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: UCKL1-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027287.1 1 2548 75
          GenBank mRNA AK128497.1 - 1 2547 75

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 647 MIR647 MIRN647,
          hsa-mir-647
          -10.7kb Anti-sense Chr. 20 - 63942631 - 63942726 on NCBI Build GRCh38
          Downstream zinc finger protein 512B ZNF512B GM632 136.0 Anti-sense Chr. 20 - 63956703 - 63969870 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000008099.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000530.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 825487 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 98224 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3356004 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_5099 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_5100 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 10909 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3356004 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3356004 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267888 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31919994 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267909 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267908 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31919999 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267878 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267890 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31919990 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31919972 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17212382 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267876 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17212381 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267870 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267894 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267896 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17212410 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267884 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267871 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267882 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267900 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31919967 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267886 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267903 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267892 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267868 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267889 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267869 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267893 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267875 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31919974 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31919997 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31919970 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31919993 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267897 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267906 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31919965 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31919952 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267886 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267868 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267906 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31919965 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31919999 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267893 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267888 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267871 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17212410 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31919972 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267897 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267900 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267892 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31919952 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31919967 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17212382 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31919970 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267870 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31919990 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267884 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267878 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31919993 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31919974 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267896 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267876 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31919994 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267875 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267890 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267869 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267889 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267894 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17212381 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267903 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267882 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31919997 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267909 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11267908 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267953 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267848 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267868 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267890 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267877 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267957 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267902 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267867 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17212365 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267905 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267881 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267841 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17212363 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267950 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267837 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267915 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17212405 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267855 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17212400 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267854 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267833 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267889 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267970 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267912 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267835 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267870 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267853 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267862 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267872 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17212444 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267954 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17212439 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267874 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17212385 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267966 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267858 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267914 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267866 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267894 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267964 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267908 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267899 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267896 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267863 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267856 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267887 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267845 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17212381 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267850 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267906 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267913 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17212384 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17212403 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267956 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267882 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267859 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267839 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267962 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267958 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267892 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11267960 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03549992 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR20007580.hg.1 TC20000530.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20007580.hg.1 NR_027287.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008099.hg.1 ENST00000623569.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008099.hg.1 ENST00000623569.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008099.hg.1 ENST00000623569.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008099.hg.1 NR_027287.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008099.hg.1 NR_027287.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20007581.hg.1 TC20000530.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20007581.hg.1 NR_027287.2 Refseq
          Affymetrix Human Clariom D Assay PSR2000156763.hg.1 ENST00000623569.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000156763.hg.1 NR_027287.2 Refseq
          Affymetrix Human Clariom D Assay PSR2000156763.hg.1 TC2000008099.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000156763.hg.1 NR_027287.2 Refseq
          Affymetrix Human Clariom D Assay PSR2000156763.hg.1 ENST00000623569.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000156765.hg.1 ENST00000623569.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000156765.hg.1 NR_027287.2 Refseq
          Affymetrix Human Clariom D Assay PSR2000156765.hg.1 TC2000008099.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000156765.hg.1 ENST00000623569.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000156765.hg.1 NR_027287.2 Refseq
          Affymetrix Human Clariom D Assay TC2000008099.hg.1 ENST00000623569.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000008099.hg.1 ENST00000623569.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000008099.hg.1 ENST00000623569.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000008099.hg.1 NR_027287.2 Refseq
          Affymetrix Human Clariom D Assay TC2000008099.hg.1 NR_027287.2 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A018594 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A018594 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 123640 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0