Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03673930_s1
          See other LOC283887 GE Assays ›
          Gene Symbol:
          LOC283887
          Gene Name:
          uncharacterized LOC283887
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 25066887 - 25068955 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03673930_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC283887
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_135196.1 1 401 93
          NR_135197.1 1 401 93
          GenBank mRNA AK093382.1 - 1 250 93
          BC023651.2 - 1 351 93
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA HY020860.1 - 1 399 Chr. 16 - 25066889 - 25067401 [+] on Build GRCh38 93
          GenBankmRNA BM552824.1 - 1 365 Chr. 16 - 25066935 - 25073435 [+] on Build GRCh38 93
          GenBankmRNA HY010984.1 - 1 261 Chr. 16 - 25067027 - 25067528 [+] on Build GRCh38 93
          GenBankmRNA BE390632.1 - 1 332 Chr. 16 - 25066956 - 25067601 [+] on Build GRCh38 93
          GenBankmRNA DB034016.1 - 1 250 Chr. 16 - 25067038 - 25067604 [+] on Build GRCh38 93
          GenBankmRNA HY036216.1 - 1 399 Chr. 16 - 25066889 - 25067364 [+] on Build GRCh38 93

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream leucine carboxyl methyltransferase 1 pseudogene LOC554206 -34.2kb Sense Chr. 16 - 25031741 - 25032777 on NCBI Build GRCh38
          Downstream LCMT1 antisense RNA 1 LCMT1-AS1 32.9kb Anti-sense Chr. 16 - 25100564 - 25111555 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30493041 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30493080 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30493070 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30493025 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968657 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968663 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30493043 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30493055 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30493024 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30493051 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30493083 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30493034 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968694 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30493059 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30493062 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30493060 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968696 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16870335 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30493071 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968665 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30493054 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968691 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16870328 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30493019 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968662 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968673 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30493065 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30493079 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968664 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30493023 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968671 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30493061 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968680 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968667 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968666 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30493072 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968677 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968688 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968675 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968656 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30493018 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968702 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968672 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968689 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 142185 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 42065 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 956823 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 308485 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 302551 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3677061 - -
          Agilent Human Genome CGH Microarray 44B A_14_P129501 - -
          Agilent Human Genome CGH Microarray 44A A_14_P129501 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 3827 - -
          Agilent Human Genome CGH + SNP Microarray A_14_P129501 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3677061 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P200490 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P129501 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P129501 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P129501 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 49769 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 2556 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P129501 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P129501 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1564373_PM_a_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1556709_PM_a_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1600007271.hg.1 ENST00000571219.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007271.hg.1 ENST00000571219.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007271.hg.1 ENST00000571219.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007271.hg.1 ENST00000571219.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007271.hg.1 ENST00000571219.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007271.hg.1 ENST00000571219.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007271.hg.1 ENST00000571219.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007271.hg.1 ENST00000571219.1 Ensembl
          Human Genome U133 Plus 2.0 Array 1556709_PM_a_at - -
          Human Genome U133 Plus 2.0 Array 1556709_a_at - -
          Human Genome U133 Plus 2.0 Array 1564373_a_at - -
          Human Genome U133 Plus 2.0 Array 1564373_PM_a_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556709_a_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564373_a_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556709_a_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564373_a_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556709_a_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564373_a_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564373_a_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556709_a_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564373_a_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564373_a_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556709_a_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556709_a_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564373_a_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556709_a_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556709_a_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564373_a_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556709_a_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564373_a_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556709_a_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564373_a_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564373_a_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556709_a_at8 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 710345 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1676545 BC023651.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1676545 AK093382.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1676545 BC023651.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1676545 AK093382.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1676545 BC023651.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1676545 AK093382.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1676545 BC023651.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1676545 AK093382.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1676545 BC023651.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1676545 AK093382.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0