Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03676048_s1
          See other LOC554206 GE Assays ›
          Gene Symbol:
          LOC554206
          Gene Name:
          leucine carboxyl methyltransferase 1 pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 25031741 - 25032777 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03676048_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC554206
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038379.1 1 51 91
          GenBank mRNA BC029609.1 - 1 48 91
          DB115487.1 - 1 51 91
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BI600002.1 - 1 53 Chr. 16 - 25031744 - 25032501 [+] on Build GRCh38 91

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream thyroid hormone receptor interactor 11 pseudogene LOC100421171 -9.4kb Sense Chr. 16 - 25015978 - 25022380 on NCBI Build GRCh38
          Downstream uncharacterized LOC283887 LOC283887 34.3kb Sense Chr. 16 - 25066887 - 25068955 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600007268.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC16000266.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968595 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968618 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30492988 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968601 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968606 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30492956 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968603 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968610 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30492947 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30492984 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968626 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30492967 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968628 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30492966 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968608 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968612 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968624 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968633 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30492983 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968607 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968632 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968600 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968609 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30492965 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968627 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30492982 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30492970 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30492981 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968611 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968623 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968597 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968614 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968591 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968592 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968605 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968625 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30492968 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968621 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968620 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09968599 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 2079 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 42717 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 142908 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 877097 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 474077 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 276 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P434040 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 34218 - -
          Agilent Human Genome CGH Microarray 44B A_14_P124057 - -
          Agilent Human Genome CGH Microarray 44A A_14_P124057 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 2275 - -
          Agilent Human Genome CGH + SNP Microarray A_14_P124057 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3294966 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P434040 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P124057 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P124057 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P124057 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P434040 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 156433 AF085481.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 156433 AK026943.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 156433 AK290378.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 156433 BC016475.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 156433 NM_004657.4 refseq_mrna_human_NM
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 97153 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P124057 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03129520 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P124057 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7994236 - -
          Affymetrix Human Gene 1.1 ST Array 7994235 - -
          Affymetrix HT HG-U133+ PM Array Plate 1554187_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1554185_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR16003989.hg.1 NR_038379.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16003989.hg.1 TC16000266.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1600007268.hg.1 ENST00000575694.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007268.hg.1 ENST00000575694.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007268.hg.1 ENST00000619106.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007268.hg.1 ENST00000619106.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007268.hg.1 ENST00000619106.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007268.hg.1 NR_038379.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007268.hg.1 NR_038379.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007268.hg.1 NR_038379.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600152373.hg.1 ENST00000575694.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600152373.hg.1 ENST00000619106.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600152373.hg.1 NR_038379.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600152373.hg.1 TC1600007268.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600152373.hg.1 ENST00000575694.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600152373.hg.1 ENST00000619106.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600152373.hg.1 NR_038379.1 Refseq
          Affymetrix Human Clariom D Assay TC1600007268.hg.1 ENST00000575694.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007268.hg.1 ENST00000575694.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007268.hg.1 ENST00000619106.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007268.hg.1 ENST00000619106.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007268.hg.1 ENST00000619106.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007268.hg.1 NR_038379.1 Refseq
          Affymetrix Human Clariom D Assay TC1600007268.hg.1 NR_038379.1 Refseq
          Affymetrix Human Clariom D Assay TC1600007268.hg.1 NR_038379.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600152374.hg.1 ENST00000619106.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600152374.hg.1 NR_038379.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600152374.hg.1 TC1600007268.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600152374.hg.1 ENST00000619106.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600152374.hg.1 NR_038379.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600152372.hg.1 ENST00000619106.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600152372.hg.1 NR_038379.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600152372.hg.1 TC1600007268.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600152372.hg.1 ENST00000619106.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600152372.hg.1 NR_038379.1 Refseq
          Human Genome U133 Plus 2.0 Array 1554185_PM_at - -
          Human Genome U133 Plus 2.0 Array 1554187_PM_at - -
          Human Genome U133 Plus 2.0 Array 1554187_at - -
          Human Genome U133 Plus 2.0 Array 1554185_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554187_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554187_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554185_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554187_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554187_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554187_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554185_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554185_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554187_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554185_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554185_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554187_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554185_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554185_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554187_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554185_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554187_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554187_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554185_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554185_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554187_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554185_at7 - -
          Human Genome U95 Set 92033_at - -
          Human Genome U95 Set 92033_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A054402 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A054402 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 694208 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1690036 BC029609.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1690036 NR_038379.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1690036 AI553759.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1690036 AI696363.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1690036 AK097891.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1754134 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1754134 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1690036 BC029609.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1690036 NR_038379.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1690036 AI553759.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1690036 AI696363.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1690036 AK097891.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix