Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03678257_s1
          See other MTVR2 GE Assays ›
          Gene Symbol:
          MTVR2
          Gene Name:
          mouse mammary tumor virus receptor homolog 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 56884102 - 56884913 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03678257_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MTVR2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027025.1 1 50 95
          GenBank mRNA AA191116.1 - 1 50 95

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream diacylglycerol kinase epsilon DGKE AHUS7,
          DAGK5,
          DAGK6,
          DGK,
          NPHS7
          -14.5kb Anti-sense Chr. 17 - 56834099 - 56869565 on NCBI Build GRCh38
          Downstream tripartite motif containing 25 TRIM25 EFP,
          RNF147,
          Z147,
          ZNF147
          3.0kb Sense Chr. 17 - 56887909 - 56914048 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700011207.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001715.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3332302 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4940 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4941 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 2412 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3332302 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3332302 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964560 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964565 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964580 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10386240 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964581 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964570 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964583 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964564 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10386232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16998913 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964571 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10386239 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964579 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964582 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964578 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10386231 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10386241 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964562 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964558 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964583 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964579 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10386241 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964580 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964570 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10386232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964558 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10386231 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964565 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10386239 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16998913 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964578 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964582 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964560 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964571 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10386240 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964562 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964564 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30964581 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10386213 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10386231 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10386226 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10386205 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10386240 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10386242 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10386220 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10386212 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10386237 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16998913 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10386229 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10386215 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16998918 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10386239 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10386224 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10386207 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10386245 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16998912 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10386204 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10386247 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10386218 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10386235 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10386210 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1700011207.hg.1 ENST00000571053.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011207.hg.1 NR_027025.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011207.hg.1 NR_027025.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011207.hg.1 NR_027025.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17023010.hg.1 NR_027025.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17023010.hg.1 TC17001715.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1700011207.hg.1 ENST00000571053.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011207.hg.1 NR_027025.1 Refseq
          Affymetrix Human Clariom D Assay TC1700011207.hg.1 NR_027025.1 Refseq
          Affymetrix Human Clariom D Assay TC1700011207.hg.1 NR_027025.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700190042.hg.1 ENST00000571053.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190042.hg.1 NR_027025.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700190042.hg.1 TC1700011207.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700190042.hg.1 NR_027025.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700190042.hg.1 ENST00000571053.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700190041.hg.1 NR_027025.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700190041.hg.1 TC1700011207.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700190041.hg.1 NR_027025.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700190043.hg.1 NR_027025.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700190043.hg.1 TC1700011207.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700190043.hg.1 NR_027025.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008605 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008605 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0