Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03678482_g1
          See other DKFZP779M0652 GE Assays ›
          Gene Symbol:
          DKFZp779M0652
          Gene Name:
          uncharacterized DKFZp779M0652
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 45771432 - 45772358 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03678482_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DKFZp779M0652
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027134.1 1 - 2 281 102
          GenBank mRNA BC126374.1 - 1 - 2 211 102
          BC126376.1 - 1 - 2 211 102
          BX640618.1 - 1 - 2 281 102
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CR748499.1 - 1 - 2 246 Chr. 11 - 45771467 - 45772115 [+] on Build GRCh38 102
          GenBankmRNA BX499991.1 - 1 - 2 Chr. 11 - 45771543 - 45772358 [-] on Build GRCh38 102
          GenBankmRNA AI742863.1 - 1 - 2 Chr. 11 - 45771563 - 45772358 [-] on Build GRCh38 102
          GenBankmRNA AI803005.1 - 1 - 2 Chr. 11 - 45771599 - 45772358 [-] on Build GRCh38 102
          GenBankmRNA BX111696.1 - 1 - 2 246 Chr. 11 - 45771467 - 45772210 [+] on Build GRCh38 102
          GenBankmRNA AI928511.1 - 1 - 2 Chr. 11 - 45771543 - 45772358 [-] on Build GRCh38 102
          GenBankmRNA AW204704.1 - 1 - 2 Chr. 11 - 45771578 - 45772358 [-] on Build GRCh38 102
          GenBankmRNA CK299586.1 - 1 - 2 Chr. 11 - 45771477 - 45772358 [-] on Build GRCh38 102
          GenBankmRNA AJ710030.1 - 2 - 3 129 Chr. 11 - 45771427 - 45772355 [+] on Build GRCh38 102
          GenBankmRNA HY011459.1 - 1 - 2 257 Chr. 11 - 45771456 - 45772242 [+] on Build GRCh38 102
          GenBankmRNA AI798867.1 - 1 - 2 Chr. 11 - 45771598 - 45772355 [-] on Build GRCh38 102
          GenBankmRNA HY047753.1 - 1 - 2 252 Chr. 11 - 45771461 - 45772194 [+] on Build GRCh38 102
          GenBankmRNA BE348376.1 - 1 - 2 Chr. 11 - 45771504 - 45772357 [-] on Build GRCh38 102
          GenBankmRNA AI743142.1 - 1 - 2 Chr. 11 - 45771552 - 45772358 [-] on Build GRCh38 102
          GenBankmRNA BM919211.1 - 2 - 3 146 Chr. 11 - 45771429 - 45772357 [+] on Build GRCh38 102
          GenBankmRNA BQ189459.1 - 1 - 2 246 Chr. 11 - 45771467 - 45772280 [+] on Build GRCh38 102

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC100507384 LOC100507384 -46.9kb Sense Chr. 11 - 45722308 - 45724555 on NCBI Build GRCh38
          Downstream solute carrier family 35 member C1 SLC35C1 CDG2C,
          FUCT1
          31.7kb Sense Chr. 11 - 45804072 - 45813016 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100007425.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11000386.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28194884 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07793426 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28194894 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07793415 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16441424 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28194915 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07793462 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07793411 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07793410 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28194928 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07793413 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07793463 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07793457 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28194887 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07793442 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07793435 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07793439 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28194899 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28194892 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07793436 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07793437 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28194924 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07793438 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07793458 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07793421 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07793451 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07793418 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07793446 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28194922 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28194914 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07793424 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28194896 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28194897 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28194930 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07793444 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07793471 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28194934 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28194929 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28194919 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28194918 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16441423 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07793412 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28194927 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28194905 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28194931 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P189790 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P189790 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 43073 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 19327 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P189790 - -
          SurePrint G3 Human GE Microarray G4851A A_32_P189790 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793425 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793410 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28194897 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28194896 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28194875 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793429 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793426 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28194894 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793421 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793415 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793413 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793436 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793437 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793414 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793424 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793411 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28194892 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28194884 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28194899 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28194887 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16441423 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793435 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28194886 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793418 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793435 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28194892 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793425 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793437 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28194899 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28194896 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793429 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793411 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793413 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793418 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793421 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793415 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28194897 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16441423 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793414 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28194884 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793426 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28194894 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793436 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793424 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28194875 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28194886 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28194887 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07793410 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P189790 - -
          Agilent Human 1B Microarray G4111A A_32_P189790 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793470 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16441430 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793457 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793440 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793449 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793426 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793451 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793434 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793436 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793418 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16441422 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793452 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793424 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793429 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793420 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793463 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793471 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793467 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793460 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793421 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793410 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793431 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16441425 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793423 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793447 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793428 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793438 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16441423 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793443 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793413 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793414 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07793465 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16441429 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7939589 - -
          Affymetrix Human Gene 1.0 ST Array 7939588 - -
          Affymetrix Human Gene 1.1 ST Array 7939587 - -
          Affymetrix HT HG-U133+ PM Array Plate 230115_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1100007425.hg.1 ENST00000378779.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007425.hg.1 ENST00000378779.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007425.hg.1 NR_027134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007425.hg.1 NR_027134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11004778.hg.1 ENST00000378779.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11004778.hg.1 NR_027134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11004778.hg.1 TC11000386.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR11004779.hg.1 ENST00000378779.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11004779.hg.1 NR_027134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11004779.hg.1 TC11000386.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1100152498.hg.1 ENST00000378779.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100152498.hg.1 NR_027134.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100152498.hg.1 TC1100007425.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100152498.hg.1 NR_027134.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100152498.hg.1 ENST00000378779.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100152499.hg.1 ENST00000378779.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100152499.hg.1 NR_027134.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100152499.hg.1 TC1100007425.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100152499.hg.1 ENST00000378779.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100152499.hg.1 NR_027134.1 Refseq
          Affymetrix Human Clariom D Assay TC1100007425.hg.1 ENST00000378779.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100007425.hg.1 ENST00000378779.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100007425.hg.1 NR_027134.1 Refseq
          Affymetrix Human Clariom D Assay TC1100007425.hg.1 NR_027134.1 Refseq
          Human Genome U133 Plus 2.0 Array 230115_at - -
          Human Genome U133 Plus 2.0 Array 230115_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230115_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230115_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230115_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230115_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230115_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230115_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230115_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230115_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230115_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230115_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230115_at1 - -
          Human Genome U95 Set 77048_at - -
          Human Genome U95 Set 77048_PM_at - -
          Human Genome U133 Set 230115_at - -
          Human Genome U133 Set 230115_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046319 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046319 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3188058 NR_027134.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3188058 BC126374.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3188058 BC126376.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3188058 BX640618.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0