Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03678782_g1
          See other FLVCR1-AS1 GE Assays ›
          Gene Symbol:
          FLVCR1-AS1
          Gene Name:
          FLVCR1 antisense RNA 1 (head to head)
          Gene Aliases:
          LQK1, NCRNA00292
          Chromosome Location:
          Chr.1: 212856604 - 212858138 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03678782_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FLVCR1-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027285.1 1 - 2 378 90
          GenBank mRNA AY030238.1 - 1 - 2 317 90
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DN996207.1 - 1 - 2 356 Chr. 1 - 212856919 - 212858112 [-] on Build GRCh38 90
          GenBankmRNA AV725773.1 - 1 - 2 197 Chr. 1 - 212856891 - 212857957 [-] on Build GRCh38 90
          GenBankmRNA BE795469.1 - 1 - 2 310 Chr. 1 - 212856705 - 212858070 [-] on Build GRCh38 90
          GenBankmRNA AV726178.1 - 1 - 2 197 Chr. 1 - 212856633 - 212857957 [-] on Build GRCh38 90
          GenBankmRNA AV727128.2 - 1 - 2 197 Chr. 1 - 212856859 - 212857957 [-] on Build GRCh38 90
          GenBankmRNA AV726293.1 - 1 - 2 197 Chr. 1 - 212856865 - 212857957 [-] on Build GRCh38 90
          GenBankmRNA CD246652.1 - 1 - 2 348 Chr. 1 - 212856701 - 212858091 [-] on Build GRCh38 90
          GenBankmRNA AV726295.1 - 1 - 2 197 Chr. 1 - 212856865 - 212857957 [-] on Build GRCh38 90
          GenBankmRNA BF686299.1 - 1 - 2 162 Chr. 1 - 212856632 - 212857911 [-] on Build GRCh38 90
          GenBankmRNA AV727007.1 - 1 - 2 197 Chr. 1 - 212856892 - 212857957 [-] on Build GRCh38 90

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream spermatogenesis associated 45 SPATA45 C1orf227 -9.0kb Sense Chr. 1 - 212830141 - 212847649 on NCBI Build GRCh38
          Downstream feline leukemia virus subgroup C cellular receptor 1 FLVCR1 AXPC1,
          FLVCR,
          MFSD7B,
          PCA,
          PCARP
          117.0 Anti-sense Chr. 1 - 212858255 - 212899363 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100017262.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01003812.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3299811 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P190404 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 15669 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 29509 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3299811 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3299811 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20843338 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15172085 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00780232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00780227 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00780229 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00780230 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00780230 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00780229 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20843338 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00780227 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15172085 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00780232 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P190404 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P190404 - -
          Agilent Human 1B Microarray G4111A A_32_P190408 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15172085 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00780232 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00780205 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00780225 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00780207 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00780202 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00780204 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00780227 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00780229 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7924188 - -
          Affymetrix Human Gene 1.0 ST Array 7924189 - -
          Affymetrix Human Gene 1.0 ST Array 7924186 - -
          Affymetrix Human Gene 1.0 ST Array 7924187 - -
          Affymetrix Human Gene 1.1 ST Array 7924184 - -
          Affymetrix HT HG-U133+ PM Array Plate 235126_PM_at - -
          Affymetrix Human Genome U219 Array 11744731_at - -
          Affymetrix Human Genome U219 Array 11744731_PM_at - -
          Affymetrix Human Genome U219 Array 11758579_PM_s_at - -
          Affymetrix Human Genome U219 Array 11758579_s_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000356684.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000356684.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000356684.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000356684.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000424044.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000424044.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 NR_027285.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 NR_027285.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 NR_027285.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 NR_027285.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 NR_027286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 NR_027286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017262.hg.1 NR_027286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01058235.hg.1 NR_027285.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01058235.hg.1 TC01003812.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000356684.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000356684.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000356684.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000356684.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000424044.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000424044.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 NR_027285.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 NR_027285.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 NR_027285.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 NR_027285.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 NR_027286.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 NR_027286.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017262.hg.1 NR_027286.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100230669.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100230669.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100230669.hg.1 NR_027285.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100230669.hg.1 TC0100017262.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100230669.hg.1 NR_027285.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100230669.hg.1 ENST00000426161.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100230669.hg.1 ENST00000426161.1 Ensembl
          Human Genome U133 Plus 2.0 Array 235126_at - -
          Human Genome U133 Plus 2.0 Array 235126_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235126_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235126_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235126_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235126_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235126_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235126_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235126_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235126_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235126_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235126_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235126_at5 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A010932 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A007495 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A007495 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A010932 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1750368 NR_027285.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1750368 NR_027286.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1750368 AY030238.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1750368 AY030239.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1668771 - -
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1683183 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1683183 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1750368 NR_027285.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1750368 NR_027286.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1750368 AY030238.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1750368 AY030239.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1668771 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1750368 NR_027285.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1750368 NR_027286.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1750368 AY030238.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1750368 AY030239.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1668771 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1782178 NR_027285.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1782178 AY030238.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1668771 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1750368 NR_027285.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1750368 NR_027286.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1750368 AY030238.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1750368 AY030239.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1782178 NR_027285.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1782178 AY030238.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-8696d955b-mh7vc:80/100.66.135.11:80.
          git-commit: b6496d4449ebd8b46e03b29da6eff4841d0bfc13
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline