Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03680813_m1
          See other LOC729609 GE Assays ›
          Gene Symbol:
          LOC729609
          Gene Name:
          uncharacterized LOC729609
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 19986817 - 19989779 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03680813_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC729609
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_024440.1 1 - 2 93 112

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream chromosome X open reading frame 23 CXorf23 Sense Chr. X - 19912860 - 19991040 on NCBI Build GRCh38
          Downstream MAP7 domain containing 2 MAP7D2 16.9kb Sense Chr. X - 20006713 - 20116996 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00009212.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X000906.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627737 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32333010 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32333006 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627742 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627727 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627732 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627707 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627752 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32332981 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32333011 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627751 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627731 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627719 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627726 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627755 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32333015 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627704 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627754 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32332980 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32332976 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627748 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32332978 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627725 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627711 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32332974 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32333012 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627747 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627738 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32333002 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32333007 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627756 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32332987 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32333003 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627758 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32332986 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627721 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32332979 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32332971 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627734 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627728 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627753 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11627759 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 125880 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 18507 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 623687 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 738697 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3410831 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4566 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4565 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 42052 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P21403088 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P21403088 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3410831 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3410831 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627734 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32333006 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627758 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627728 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32332986 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627752 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627748 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627721 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627760 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627759 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32333011 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627753 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32333010 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627719 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627751 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32332978 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32333016 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32333003 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32333002 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32333012 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627731 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32333007 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32332987 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627717 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32332982 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32332981 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32333025 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627743 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627756 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627720 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32333028 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627747 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627726 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627737 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32332979 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627732 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627754 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32333026 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627726 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627751 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627737 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32332979 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627756 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32333025 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627759 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627731 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627728 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627758 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627753 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627743 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627734 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627747 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32333010 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32333011 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32333012 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32333002 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32333026 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32333006 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32332981 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32332986 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627717 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627720 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32332982 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32333028 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627721 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32333003 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32332978 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627732 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627752 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627748 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32332987 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32333016 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627719 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32333007 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627760 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11627754 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P21403088 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P21403088 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 96499 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 90417 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21403088 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03666794 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627716 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17324337 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627758 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627728 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627708 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627762 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627706 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627736 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627740 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627750 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627761 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627723 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627743 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627703 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17324336 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627719 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17324334 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627726 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627752 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17324332 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627746 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627745 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627765 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627713 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627754 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627732 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627748 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627734 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627710 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627705 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627738 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11627756 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03666794 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P21403088 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR0X012229.hg.1 NR_024440.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X012229.hg.1 TC0X000906.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR0X012230.hg.1 ENST00000500992.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X012230.hg.1 NR_024440.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X012230.hg.1 TC0X000906.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR0X012232.hg.1 ENST00000500992.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X012232.hg.1 NR_024440.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X012232.hg.1 TC0X000906.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0X00163292.hg.1 NR_024440.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00163292.hg.1 TC0X00009212.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00163292.hg.1 NR_024440.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00163291.hg.1 NR_024440.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00163291.hg.1 TC0X00009212.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00163291.hg.1 NR_024440.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00163294.hg.1 NR_024440.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00163294.hg.1 TC0X00009212.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00163294.hg.1 NR_024440.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A007309 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A007309 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3249715 NR_024440.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3249715 AW976820.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3249715 AY092822.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3249715 NR_024440.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3249715 AW976820.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3249715 AY092822.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3249715 NR_024440.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3249715 AW976820.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3249715 AY092822.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3249715 NR_024440.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3249715 AW976820.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3249715 AY092822.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0