Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03681728_s1
          See other FAM225A GE Assays ›
          Gene Symbol:
          FAM225A
          Gene Name:
          family with sequence similarity 225 member A (non-protein coding)
          Gene Aliases:
          C9orf109, LINC00256A, NCRNA00256A
          Chromosome Location:
          Chr.9: 113112896 - 113119846 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03681728_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FAM225A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_024366.1 3 5356 96
          GenBank mRNA AK127516.1 - 1 4342 96
          AL832752.1 - 3 5179 96
          BX647840.1 - 2 5583 96
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CA306976.1 - 1 Chr. 9 - 113119081 - 113119848 [-] on Build GRCh38 96
          GenBankmRNA DB336653.1 - 1 Chr. 9 - 113119259 - 113119844 [-] on Build GRCh38 96
          GenBankmRNA BU627090.1 - 1 Chr. 9 - 113119208 - 113119848 [-] on Build GRCh38 96
          GenBankmRNA BG164717.1 - 1 513 Chr. 9 - 113118888 - 113119677 [+] on Build GRCh38 96
          GenBankmRNA DB304829.1 - 1 Chr. 9 - 113119296 - 113119844 [-] on Build GRCh38 96

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream family with sequence similarity 225 member B (non-protein coding) FAM225B C9orf110,
          LINC00256B,
          NCRNA00256B
          -2.7kb Anti-sense Chr. 9 - 113104723 - 113111677 on NCBI Build GRCh38
          Downstream solute carrier family 31 member 2 SLC31A2 COPT2,
          CTR2,
          hCTR2
          31.1kb Sense Chr. 9 - 113150958 - 113164142 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900008481.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09000570.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CNV Microarray 2x400K 244517 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P687326 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 15037 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 28064 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P687326 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P687326 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27249816 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27249767 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27249768 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06905782 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27249819 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27249813 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06905774 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27249768 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27249813 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06905774 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06905782 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27249767 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27249816 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27249819 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P687326 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905767 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16255007 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905768 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16255129 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905780 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905791 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905761 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905808 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905762 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905778 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905759 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905749 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905802 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905803 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905806 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905757 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905760 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905774 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16255130 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905794 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905798 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905781 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905789 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905783 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905800 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905787 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905786 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905765 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905810 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905773 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905776 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905770 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905755 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06905752 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR09006320.hg.1 ENST00000434051.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09006320.hg.1 NR_024366.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09006320.hg.1 TC09000570.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0900008481.hg.1 ENST00000434051.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008481.hg.1 ENST00000434051.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008481.hg.1 ENST00000434051.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008481.hg.1 ENST00000434051.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008481.hg.1 ENST00000434051.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008481.hg.1 ENST00000453010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008481.hg.1 ENST00000453010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008481.hg.1 ENST00000453010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008481.hg.1 NR_024366.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008481.hg.1 NR_024366.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008481.hg.1 NR_024366.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008481.hg.1 NR_024366.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09006322.hg.1 ENST00000434051.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09006322.hg.1 NR_024366.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09006322.hg.1 TC09000570.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09006319.hg.1 NR_024366.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09006319.hg.1 TC09000570.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0900011250.hg.1 ENST00000423546.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011250.hg.1 ENST00000423546.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011250.hg.1 ENST00000423546.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011250.hg.1 ENST00000423546.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011250.hg.1 ENST00000439875.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011250.hg.1 ENST00000439875.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011250.hg.1 ENST00000439875.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011250.hg.1 ENST00000439875.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011250.hg.1 ENST00000439875.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011250.hg.1 ENST00000439875.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011250.hg.1 NR_024376.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011250.hg.1 NR_024376.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011250.hg.1 NR_024376.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011250.hg.1 NR_024376.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011250.hg.1 NR_024376.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09006323.hg.1 ENST00000434051.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09006323.hg.1 NR_024366.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09006323.hg.1 TC09000570.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0900156594.hg.1 NR_024366.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900156594.hg.1 TC0900008481.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900156594.hg.1 NR_024366.1 Refseq
          Affymetrix Human Clariom D Assay TC0900011250.hg.1 ENST00000423546.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011250.hg.1 ENST00000423546.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011250.hg.1 ENST00000423546.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011250.hg.1 ENST00000423546.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011250.hg.1 ENST00000439875.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011250.hg.1 ENST00000439875.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011250.hg.1 ENST00000439875.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011250.hg.1 ENST00000439875.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011250.hg.1 ENST00000439875.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011250.hg.1 ENST00000439875.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011250.hg.1 NR_024376.1 Refseq
          Affymetrix Human Clariom D Assay TC0900011250.hg.1 NR_024376.1 Refseq
          Affymetrix Human Clariom D Assay TC0900011250.hg.1 NR_024376.1 Refseq
          Affymetrix Human Clariom D Assay TC0900011250.hg.1 NR_024376.1 Refseq
          Affymetrix Human Clariom D Assay TC0900011250.hg.1 NR_024376.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900156597.hg.1 ENST00000434051.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900156597.hg.1 ENST00000453010.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900156597.hg.1 NR_024366.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900156597.hg.1 TC0900008481.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900156597.hg.1 ENST00000434051.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900156597.hg.1 ENST00000453010.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900156597.hg.1 NR_024366.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900156598.hg.1 ENST00000434051.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900156598.hg.1 ENST00000453010.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900156598.hg.1 NR_024366.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900156598.hg.1 TC0900008481.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900156598.hg.1 ENST00000434051.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900156598.hg.1 ENST00000453010.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900156598.hg.1 NR_024366.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900156595.hg.1 ENST00000434051.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900156595.hg.1 ENST00000453010.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900156595.hg.1 NR_024366.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900156595.hg.1 TC0900008481.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900156595.hg.1 ENST00000434051.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900156595.hg.1 ENST00000453010.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900156595.hg.1 NR_024366.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008481.hg.1 ENST00000434051.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008481.hg.1 ENST00000434051.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008481.hg.1 ENST00000434051.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008481.hg.1 ENST00000434051.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008481.hg.1 ENST00000434051.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008481.hg.1 ENST00000453010.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008481.hg.1 ENST00000453010.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008481.hg.1 ENST00000453010.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008481.hg.1 NR_024366.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008481.hg.1 NR_024366.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008481.hg.1 NR_024366.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008481.hg.1 NR_024366.1 Refseq
          Human Genome U133 Plus 2.0 Array 229059_at - -
          Human Genome U133 Plus 2.0 Array 229059_PM_at - -
          Human Genome U95 Set 52878_at - -
          Human Genome U95 Set 52878_PM_at - -
          Human Genome U133 Set 229059_PM_at - -
          Human Genome U133 Set 229059_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A039123 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A037925 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A039123 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A037925 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1695485 BX647840.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1695485 AK127516.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1695485 AL832752.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1695485 NR_024366.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1695485 BX647840.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1695485 AK127516.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1695485 AL832752.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1695485 NR_024366.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1695485 BX647840.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1695485 AK127516.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1695485 AL832752.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1695485 NR_024366.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1695485 BX647840.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1695485 AK127516.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1695485 AL832752.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1695485 NR_024366.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0