Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008586.hg.1 |
ENST00000450938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008586.hg.1 |
ENST00000450938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008586.hg.1 |
ENST00000450938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008586.hg.1 |
ENST00000450938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008586.hg.1 |
ENST00000450938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008586.hg.1 |
ENST00000450938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008586.hg.1 |
NR_109806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008586.hg.1 |
NR_109806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008586.hg.1 |
NR_109806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008586.hg.1 |
NR_109806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008586.hg.1 |
NR_109806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008586.hg.1 |
NR_109806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900157276.hg.1 |
ENST00000450938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900157276.hg.1 |
NR_109806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900157276.hg.1 |
TC0900008586.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900157276.hg.1 |
NR_109806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900157276.hg.1 |
ENST00000450938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900157277.hg.1 |
ENST00000450938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900157277.hg.1 |
NR_109806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900157277.hg.1 |
TC0900008586.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900157277.hg.1 |
ENST00000450938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900157277.hg.1 |
NR_109806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900157278.hg.1 |
ENST00000450938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900157278.hg.1 |
NR_109806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900157278.hg.1 |
TC0900008586.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900157278.hg.1 |
ENST00000450938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900157278.hg.1 |
NR_109806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008586.hg.1 |
ENST00000450938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008586.hg.1 |
ENST00000450938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008586.hg.1 |
ENST00000450938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008586.hg.1 |
ENST00000450938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008586.hg.1 |
ENST00000450938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008586.hg.1 |
ENST00000450938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008586.hg.1 |
NR_109806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008586.hg.1 |
NR_109806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008586.hg.1 |
NR_109806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008586.hg.1 |
NR_109806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008586.hg.1 |
NR_109806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008586.hg.1 |
NR_109806.1 |
Refseq |