Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008792.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008792.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008792.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008792.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008792.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008792.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008792.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008792.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008792.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008792.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008792.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008792.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008792.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008792.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008792.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008792.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008792.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008792.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700165708.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700165708.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700165708.hg.1 |
TC1700008792.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700165708.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700165708.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700165711.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700165711.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700165711.hg.1 |
TC1700008792.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700165711.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700165711.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700165710.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700165710.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700165710.hg.1 |
TC1700008792.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700165710.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700165710.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700165707.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700165707.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700165707.hg.1 |
TC1700008792.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700165707.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700165707.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008792.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008792.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008792.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008792.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008792.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008792.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008792.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008792.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008792.hg.1 |
ENST00000419257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008792.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008792.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008792.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008792.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008792.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008792.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008792.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008792.hg.1 |
NR_110827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008792.hg.1 |
NR_110827.1 |
Refseq |