Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
ENST00000502351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
ENST00000502351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
ENST00000502351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
ENST00000502351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
ENST00000505703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
ENST00000505703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
ENST00000505703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
ENST00000505703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
ENST00000505703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008845.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010241.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010241.hg.1 |
TC05000720.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0500161102.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161102.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500161102.hg.1 |
TC0500008845.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500161102.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500161102.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
ENST00000502351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
ENST00000502351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
ENST00000502351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
ENST00000502351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
ENST00000505703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
ENST00000505703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
ENST00000505703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
ENST00000505703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
ENST00000505703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008845.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500161100.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161100.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500161100.hg.1 |
TC0500008845.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500161100.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161100.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500161101.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161101.hg.1 |
NM_005859.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500161101.hg.1 |
TC0500008845.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500161101.hg.1 |
ENST00000331327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161101.hg.1 |
NM_005859.4 |
Refseq |