Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006676.hg.1 |
ENST00000448835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006676.hg.1 |
ENST00000448835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006676.hg.1 |
ENST00000448835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006676.hg.1 |
ENST00000448835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006676.hg.1 |
ENST00000448835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006676.hg.1 |
NR_120634.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006676.hg.1 |
NR_120634.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006676.hg.1 |
NR_120634.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006676.hg.1 |
NR_120634.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006676.hg.1 |
NR_120634.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006676.hg.1 |
NR_120634.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006676.hg.1 |
NR_120634.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006676.hg.1 |
NR_120634.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006676.hg.1 |
NR_120634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000145796.hg.1 |
ENST00000448835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000145796.hg.1 |
NR_120634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000145796.hg.1 |
TC1000006676.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000145796.hg.1 |
ENST00000448835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000145796.hg.1 |
NR_120634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000145794.hg.1 |
ENST00000448835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000145794.hg.1 |
NR_120634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000145794.hg.1 |
TC1000006676.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000145794.hg.1 |
ENST00000448835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000145794.hg.1 |
NR_120634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000145797.hg.1 |
ENST00000448835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000145797.hg.1 |
NR_120634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000145797.hg.1 |
TC1000006676.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000145797.hg.1 |
ENST00000448835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000145797.hg.1 |
NR_120634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006676.hg.1 |
ENST00000448835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006676.hg.1 |
ENST00000448835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006676.hg.1 |
ENST00000448835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006676.hg.1 |
ENST00000448835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006676.hg.1 |
ENST00000448835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006676.hg.1 |
NR_120634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006676.hg.1 |
NR_120634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006676.hg.1 |
NR_120634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006676.hg.1 |
NR_120634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006676.hg.1 |
NR_120634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006676.hg.1 |
NR_120634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006676.hg.1 |
NR_120634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006676.hg.1 |
NR_120634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006676.hg.1 |
NR_120634.1 |
Refseq |