Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010650.hg.1 |
ENST00000523769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010650.hg.1 |
ENST00000523769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010650.hg.1 |
ENST00000523769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013473.hg.1 |
ENST00000604716.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013473.hg.1 |
ENST00000604716.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013473.hg.1 |
ENST00000604716.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013473.hg.1 |
ENST00000604716.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013473.hg.1 |
ENST00000604716.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013473.hg.1 |
ENST00000604716.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010650.hg.1 |
ENST00000522106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010650.hg.1 |
ENST00000523769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010650.hg.1 |
ENST00000523769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010650.hg.1 |
ENST00000523769.1 |
Ensembl |