Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007542.hg.1 |
ENST00000558050.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007542.hg.1 |
ENST00000558050.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007542.hg.1 |
ENST00000558050.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007542.hg.1 |
ENST00000558050.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007542.hg.1 |
NR_120344.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007542.hg.1 |
NR_120344.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007542.hg.1 |
NR_120344.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007542.hg.1 |
NR_120344.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007542.hg.1 |
ENST00000558050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007542.hg.1 |
ENST00000558050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007542.hg.1 |
ENST00000558050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007542.hg.1 |
ENST00000558050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007542.hg.1 |
NR_120344.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007542.hg.1 |
NR_120344.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007542.hg.1 |
NR_120344.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007542.hg.1 |
NR_120344.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500153432.hg.1 |
ENST00000558050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153432.hg.1 |
NR_120344.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500153432.hg.1 |
TC1500007542.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500153432.hg.1 |
NR_120344.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500153432.hg.1 |
ENST00000558050.1 |
Ensembl |