Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000354957.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000354957.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000354957.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000354957.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000354957.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000354957.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000354957.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000354957.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000354957.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000595418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000595418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000595418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000595418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000596690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000596690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000596690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000596690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000597882.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000597882.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000597882.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000597882.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000599530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000599530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000599530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000599530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000599530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000599671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000599671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000599671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000599671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000600738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000600738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000600738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000600738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000600738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000600738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
ENST00000600738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
NM_023074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
NM_023074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
NM_023074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
NM_023074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
NM_023074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
NM_023074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
NM_023074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
NM_023074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012019.hg.1 |
NM_023074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000354957.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000354957.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000354957.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000354957.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000354957.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000354957.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000354957.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000354957.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000354957.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000595418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000595418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000595418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000595418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000596690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000596690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000596690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000596690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000597882.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000597882.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000597882.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000597882.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000599530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000599530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000599530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000599530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000599530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000599671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000599671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000599671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000599671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000600738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000600738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000600738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000600738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000600738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000600738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
ENST00000600738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
NM_023074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
NM_023074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
NM_023074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
NM_023074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
NM_023074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
NM_023074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
NM_023074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
NM_023074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012019.hg.1 |
NM_023074.3 |
Refseq |