Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008670.hg.1 |
ENST00000422488.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008670.hg.1 |
ENST00000422488.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008670.hg.1 |
ENST00000422488.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008670.hg.1 |
ENST00000422488.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008670.hg.1 |
NR_110141.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008670.hg.1 |
NR_110141.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008670.hg.1 |
NR_110141.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008670.hg.1 |
NR_110141.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008670.hg.1 |
ENST00000422488.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008670.hg.1 |
ENST00000422488.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008670.hg.1 |
ENST00000422488.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008670.hg.1 |
ENST00000422488.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008670.hg.1 |
NR_110141.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008670.hg.1 |
NR_110141.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008670.hg.1 |
NR_110141.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008670.hg.1 |
NR_110141.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700161331.hg.1 |
ENST00000422488.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700161331.hg.1 |
NR_110141.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700161331.hg.1 |
TC0700008670.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700161331.hg.1 |
NR_110141.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700161331.hg.1 |
ENST00000422488.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700161332.hg.1 |
ENST00000422488.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700161332.hg.1 |
NR_110141.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700161332.hg.1 |
TC0700008670.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700161332.hg.1 |
ENST00000422488.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700161332.hg.1 |
NR_110141.1 |
Refseq |