Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03881062_s1
          See other LOC101926967 GE Assays ›
          Gene Symbol:
          LOC101926967
          Gene Name:
          uncharacterized LOC101926967
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 44198851 - 44223703 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03881062_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC101926967
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC050302.1 - 1 1148 61
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AI703433.1 - 1 231 Chr. 17 - 44222838 - 44223251 [+] on Build GRCh38 61
          GenBankmRNA AW016578.1 - 1 249 Chr. 17 - 44222820 - 44223207 [+] on Build GRCh38 61
          GenBankmRNA BE670190.1 - 1 236 Chr. 17 - 44222833 - 44223227 [+] on Build GRCh38 61
          GenBankmRNA BE670121.1 - 1 238 Chr. 17 - 44222831 - 44223251 [+] on Build GRCh38 61
          GenBankmRNA AI702759.1 - 1 234 Chr. 17 - 44222835 - 44223159 [+] on Build GRCh38 61
          GenBankmRNA BE222166.1 - 1 230 Chr. 17 - 44222844 - 44223162 [+] on Build GRCh38 61
          GenBankmRNA AA977662.1 - 1 231 Chr. 17 - 44222838 - 44223217 [+] on Build GRCh38 61
          GenBankmRNA CA311052.1 - 1 Chr. 17 - 44222817 - 44223284 [-] on Build GRCh38 61
          GenBankmRNA AA976120.1 - 1 211 Chr. 17 - 44222858 - 44223218 [+] on Build GRCh38 61
          GenBankmRNA AA989119.1 - 1 104 Chr. 17 - 44222965 - 44223249 [+] on Build GRCh38 61
          GenBankmRNA AI299473.1 - 1 231 Chr. 17 - 44222838 - 44223216 [+] on Build GRCh38 61
          GenBankmRNA DB331009.1 - 1 Chr. 17 - 44222728 - 44223266 [-] on Build GRCh38 61
          GenBankmRNA AI824312.1 - 1 245 Chr. 17 - 44222824 - 44223228 [+] on Build GRCh38 61
          GenBankmRNA AI274302.1 - 1 238 Chr. 17 - 44222831 - 44223277 [+] on Build GRCh38 61
          GenBankmRNA BE464732.1 - 1 230 Chr. 17 - 44222844 - 44223218 [+] on Build GRCh38 61
          GenBankmRNA AA232746.1 - 1 Chr. 17 - 44222843 - 44223284 [-] on Build GRCh38 61
          GenBankmRNA AI392831.1 - 1 242 Chr. 17 - 44222827 - 44223216 [+] on Build GRCh38 61
          GenBankmRNA AA232251.1 - 1 232 Chr. 17 - 44222837 - 44223224 [+] on Build GRCh38 61
          GenBankmRNA AA480166.1 - 1 232 Chr. 17 - 44222837 - 44223227 [+] on Build GRCh38 61
          GenBankmRNA AI222805.1 - 1 234 Chr. 17 - 44222835 - 44223308 [+] on Build GRCh38 61
          GenBankmRNA BM968756.1 - 1 Chr. 17 - 44222819 - 44223283 [-] on Build GRCh38 61
          GenBankmRNA BM969761.1 - 1 Chr. 17 - 44222817 - 44223284 [-] on Build GRCh38 61
          GenBankmRNA BE222150.1 - 1 231 Chr. 17 - 44222838 - 44223216 [+] on Build GRCh38 61

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream upstream binding transcription factor,
          RNA polymerase I
          UBTF NOR-90,
          UBF,
          UBF-1,
          UBF1,
          UBF2
          -289.0 Anti-sense Chr. 17 - 44205033 - 44221626 on NCBI Build GRCh38
          Downstream solute carrier family 4 member 1 (Diego blood group) SLC4A1 AE1,
          BND3,
          CD233,
          CHC,
          DI,
          EMPB3,
          EPB3,
          FR,
          RTA1A,
          SAO,
          SPH4,
          SW,
          WD,
          WD1,
          WR
          24.7kb Anti-sense Chr. 17 - 44248390 - 44268161 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1700007990.hg.1 ENST00000563394.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007990.hg.1 ENST00000563394.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007990.hg.1 ENST00000563394.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007990.hg.1 ENST00000588279.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007990.hg.1 ENST00000588279.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007990.hg.1 ENST00000563394.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007990.hg.1 ENST00000563394.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007990.hg.1 ENST00000563394.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007990.hg.1 ENST00000588279.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007990.hg.1 ENST00000588279.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0