Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927780 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110416.1 | 3 - 4 | 376 | 94 | ||
NR_110417.1 | 2 - 3 | 112 | 94 | |||
GenBank mRNA | AW275370.1 | - | 2 - 3 | 94 | ||
BX648502.1 | - | 3 - 4 | 378 | 94 | ||
HY039622.1 | - | 3 - 4 | 343 | 94 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | protein kinase C eta | PRKCH |
PKC-L, PKCL, PRKCL, nPKC-eta |
-9.8kb | Anti-sense | Chr. 14 - 61280953 - 61550980 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC400221 | FLJ22447 | 5.2kb | Anti-sense | Chr. 14 - 61570540 - 61654713 on NCBI Build GRCh38 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |