Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03915222_s1
          See other HYDIN GE Assays ›
          Gene Symbol:
          HYDIN
          Gene Name:
          HYDIN, axonemal central pair apparatus protein
          Gene Aliases:
          CILD5, HYDIN1, HYDIN2, PPP1R31
          Chromosome Location:
          Chr.16: 70802084 - 71230722 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03915222_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HYDIN
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001270974.2 NP_001257903.1 86 20907 96
          XM_006721206.2 XP_006721269.1 86 20801 96
          XM_011523146.2 XP_011521448.1 87 20936 96
          XM_011523147.1 XP_011521449.1 87 20903 96
          XM_011523148.1 XP_011521450.1 87 20976 96
          XM_011523151.1 XP_011521453.1 86 20833 96
          XM_011523152.1 XP_011521454.1 49 14660 96
          XM_011523155.2 XP_011521457.1 42 13661 96
          XM_017023346.1 XP_016878835.1 86 21079 96
          XM_017023347.1 XP_016878836.1 74 19318 96
          XM_017023348.1 XP_016878837.1 74 19058 96
          GenBank mRNA AK125886.1 - 2 3643 96
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DB339909.1 - 1 Chr. 16 - 70802087 - 70802636 [+] on Build GRCh38 96
          GenBankmRNA BM127786.1 - 1 Chr. 16 - 70802082 - 70802550 [+] on Build GRCh38 96
          GenBankmRNA BM981820.1 - 1 Chr. 16 - 70802082 - 70802808 [+] on Build GRCh38 96
          GenBankmRNA CK819145.1 - 1 Chr. 16 - 70802082 - 70802643 [+] on Build GRCh38 96
          GenBankmRNA BM976605.1 - 1 Chr. 16 - 70802082 - 70802837 [+] on Build GRCh38 96
          GenBankmRNA BQ185360.1 - 1 Chr. 16 - 70802086 - 70802552 [+] on Build GRCh38 96

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream Vac14,
          PIKFYVE complex component
          VAC14 ArPIKfyve,
          TAX1BP2,
          TRX
          -921.0 Sense Chr. 16 - 70687439 - 70801169 on NCBI Build GRCh38
          Downstream cap methyltransferase 2 CMTR2 AFT,
          FTSJD1,
          HMTr2,
          MTr2
          478.1kb Sense Chr. 16 - 71282300 - 71290178 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s)

          epithelial cell development
          cilium movement
          ventricular system development
          trachea development
          axonemal central apparatus assembly

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1600010777.hg.1 ENST00000574178.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010777.hg.1 ENST00000574178.1 Ensembl
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 137436 - -
          ABI Human Genome Survey Microarray Version 2.0 149923 - -
          ABI Human Genome Survey Microarray Version 2.0 167179 - -
          ABI Human Genome Survey Microarray Version 2.0 171600 - -
          ABI Human Genome Survey Microarray Version 2.0 236227 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0