Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000223273.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000223273.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000223273.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000223273.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000223273.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000223273.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000223273.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000223273.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000432096.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000432096.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000432096.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000432096.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000432096.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000432096.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000432096.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000446267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000446267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000448268.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000448268.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000448268.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000448268.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000469737.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000469737.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000474392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000474392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000474392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000474392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000485025.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000485025.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000485025.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000485025.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000485025.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
ENST00000485025.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
NM_001282446.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
NM_001282446.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
NM_001282446.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
NM_001282446.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
NM_001282446.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
NM_001282446.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
NM_001282446.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
NM_001282446.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
NM_020192.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
NM_020192.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
NM_020192.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
NM_020192.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
NM_020192.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
NM_020192.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
NM_020192.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007267.hg.1 |
NM_020192.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000223273.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000223273.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000223273.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000223273.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000223273.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000223273.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000223273.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000223273.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000432096.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000432096.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000432096.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000432096.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000432096.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000432096.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000432096.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000446267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000446267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000448268.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000448268.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000448268.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000448268.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000469737.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000469737.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000474392.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000474392.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000474392.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000474392.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000485025.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000485025.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000485025.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000485025.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000485025.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
ENST00000485025.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
NM_001282446.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
NM_001282446.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
NM_001282446.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
NM_001282446.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
NM_001282446.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
NM_001282446.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
NM_001282446.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
NM_001282446.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
NM_020192.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
NM_020192.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
NM_020192.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
NM_020192.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
NM_020192.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
NM_020192.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
NM_020192.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007267.hg.1 |
NM_020192.4 |
Refseq |