Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03928521_s1
          See other HMBOX1 GE Assays ›
          Gene Symbol:
          HMBOX1
          Gene Name:
          homeobox containing 1
          Gene Aliases:
          HNF1LA, HOT1, PBHNF
          Chromosome Location:
          Chr.8: 28890370 - 29064939 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03928521_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HMBOX1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_005273635.1 XP_005273692.1 11 15133 102
          XM_017013823.1 XP_016869312.1 11 15168 102
          XM_017013824.1 XP_016869313.1 11 14985 102
          GenBank mRNA BX648950.1 - 1 5848 102
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AA861309.1 - 1 Chr. 8 - 29064538 - 29064943 [-] on Build GRCh38 102
          GenBankmRNA BE672504.1 - 1 Chr. 8 - 29064413 - 29064938 [-] on Build GRCh38 102
          GenBankmRNA AW300741.1 - 1 Chr. 8 - 29064442 - 29064843 [-] on Build GRCh38 102
          GenBankmRNA AI922087.1 - 1 Chr. 8 - 29064492 - 29064843 [-] on Build GRCh38 102
          GenBankmRNA W73553.1 - 416 Chr. - - [-] on Build 103
          GenBankmRNA BF430944.1 - 1 Chr. 8 - 29064408 - 29064885 [-] on Build GRCh38 102
          GenBankmRNA BE644949.1 - 1 Chr. 8 - 29064467 - 29064941 [-] on Build GRCh38 102
          GenBankmRNA BG472576.1 - 1 421 Chr. 8 - 29064195 - 29064777 [+] on Build GRCh38 102
          GenBankmRNA HY265426.1 - 1 Chr. 8 - 29064455 - 29064932 [-] on Build GRCh38 102
          GenBankmRNA CA416498.1 - 1 Chr. 8 - 29064236 - 29064945 [-] on Build GRCh38 102
          GenBankmRNA AA470137.1 - 1 Chr. 8 - 29064398 - 29064887 [-] on Build GRCh38 102
          GenBankmRNA T70546.1 - Chr. - - [-] on Build 102
          GenBankmRNA CA431816.1 - 1 Chr. 8 - 29064236 - 29064945 [-] on Build GRCh38 102
          GenBankmRNA BU630670.1 - 1 Chr. 8 - 29064236 - 29064945 [-] on Build GRCh38 102
          GenBankmRNA BE503363.1 - 1 Chr. 8 - 29064490 - 29064945 [-] on Build GRCh38 102
          GenBankmRNA AI524288.1 - 1 Chr. 8 - 29064464 - 29064937 [-] on Build GRCh38 102
          GenBankmRNA AI087285.1 - 1 Chr. 8 - 29064492 - 29064941 [-] on Build GRCh38 102
          GenBankmRNA BQ722936.1 - 1 535 Chr. 8 - 29064080 - 29064921 [+] on Build GRCh38 102
          GenBankmRNA BU682805.1 - 1 Chr. 8 - 29064390 - 29064945 [-] on Build GRCh38 102
          GenBankmRNA BM976228.1 - 1 Chr. 8 - 29064446 - 29064897 [-] on Build GRCh38 102
          GenBankmRNA BQ003634.1 - 1 Chr. 8 - 29064314 - 29064945 [-] on Build GRCh38 102
          GenBankmRNA AI768761.1 - 1 Chr. 8 - 29064540 - 29064941 [-] on Build GRCh38 102
          GenBankmRNA BF111101.1 - 1 Chr. 8 - 29064389 - 29064941 [-] on Build GRCh38 102
          GenBankmRNA AW294031.1 - 1 Chr. 8 - 29064378 - 29064887 [-] on Build GRCh38 102
          GenBankmRNA AA694256.1 - 1 Chr. 8 - 29064483 - 29064945 [-] on Build GRCh38 102
          GenBankmRNA AW293788.1 - 1 Chr. 8 - 29064454 - 29064887 [-] on Build GRCh38 102
          GenBankmRNA AA385747.1 - 1 78 Chr. 8 - 29064537 - 29064852 [+] on Build GRCh38 102
          GenBankmRNA AW166178.1 - 1 Chr. 8 - 29064488 - 29064843 [-] on Build GRCh38 102
          GenBankmRNA HY078055.1 - 1 320 Chr. 8 - 29064296 - 29064834 [+] on Build GRCh38 102
          GenBankmRNA AI634672.1 - 1 Chr. 8 - 29064423 - 29064938 [-] on Build GRCh38 102
          GenBankmRNA BQ017112.1 - 1 Chr. 8 - 29064433 - 29064945 [-] on Build GRCh38 102
          GenBankmRNA AW269301.1 - 1 Chr. 8 - 29064468 - 29064935 [-] on Build GRCh38 102
          GenBankmRNA AI972534.1 - 1 Chr. 8 - 29064399 - 29064937 [-] on Build GRCh38 102

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream integrator complex subunit 9 INTS9 CPSF2L,
          INT9,
          RC74
          -168.5kb Anti-sense Chr. 8 - 28767658 - 28890226 on NCBI Build GRCh38
          Downstream kinesin family member 13B KIF13B GAKIN 2.3kb Anti-sense Chr. 8 - 29067278 - 29263384 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA-dependent DNA biosynthetic process
          transcription, DNA-templated
          positive regulation of telomere maintenance via telomerase
          positive regulation of chromatin binding
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of telomerase activity
          double-stranded telomeric DNA binding
          telomerase activity
          protein binding
          telomeric DNA binding
          protein-containing complex binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix