Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04184906_g1
          See other RPS12 GE Assays ›
          Gene Symbol:
          RPS12
          Gene Name:
          ribosomal protein S12
          Gene Aliases:
          S12
          Chromosome Location:
          Chr.6: 132814569 - 132817564 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04184906_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RPS12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001016.3 NP_001007.2 1 42 55
          GenBank mRNA AK311826.1 - 1 - 2 37 55
          BC095424.1 - 1 32 30792687 55
          BQ276878.1 - 1 - 2 45 55
          D28378.1 - 1 - 2 41 55

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600009488.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000996.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK311826.1, BC095424.1, D28378.1, NM_001016.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RPS12

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
          rRNA processing
          translation
          translational initiation
          SRP-dependent cotranslational protein targeting to membrane
          viral transcription
          RNA binding
          structural constituent of ribosome

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200012355.hg.1 ENST00000414823.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007380.hg.1 ENST00000494679.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009523.hg.1 ENST00000466297.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008203.hg.1 ENST00000442381.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006638.hg.1 ENST00000437701.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008188.hg.1 ENST00000465542.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008275.hg.1 ENST00000419579.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013564.hg.1 ENST00000440335.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06011735.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06011735.hg.1 NM_001016.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06011735.hg.1 TC06000996.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06011737.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06011737.hg.1 NM_001016.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06011737.hg.1 TC06000996.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06011738.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06011738.hg.1 NM_001016.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06011738.hg.1 TC06000996.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06011741.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06011741.hg.1 NM_001016.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06011741.hg.1 TC06000996.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 NM_001016.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 NM_001016.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 NM_001016.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 NM_001016.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009488.hg.1 NM_001016.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06011739.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06011739.hg.1 NM_001016.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06011739.hg.1 TC06000996.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0400011609.hg.1 ENST00000497190.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400010828.hg.1 ENST00000461060.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010871.hg.1 ENST00000438591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007037.hg.1 ENST00000463278.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007099.hg.1 ENST00000407932.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06011736.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06011736.hg.1 NM_001016.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06011736.hg.1 TC06000996.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0300009309.hg.1 ENST00000484182.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010304.hg.1 ENST00000399739.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008301.hg.1 ENST00000492017.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008640.hg.1 ENST00000412619.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011293.hg.1 ENST00000510537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011627.hg.1 ENST00000428553.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011627.hg.1 ENST00000428553.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010176.hg.1 ENST00000493002.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008442.hg.1 ENST00000489699.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011207.hg.1 ENST00000439728.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007099.hg.1 ENST00000407932.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 NM_001016.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 NM_001016.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 NM_001016.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 NM_001016.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009488.hg.1 NM_001016.3 Refseq
          Affymetrix Human Clariom D Assay TC1300008640.hg.1 ENST00000412619.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008442.hg.1 ENST00000489699.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600165073.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600165073.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600165073.hg.1 NM_001016.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600165073.hg.1 TC0600009488.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600165073.hg.1 NM_001016.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600165073.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600165073.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600165075.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600165075.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600165075.hg.1 NM_001016.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600165075.hg.1 TC0600009488.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600165075.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600165075.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600165075.hg.1 NM_001016.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007037.hg.1 ENST00000463278.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009309.hg.1 ENST00000484182.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010304.hg.1 ENST00000399739.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500010176.hg.1 ENST00000493002.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010871.hg.1 ENST00000438591.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007380.hg.1 ENST00000494679.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200012355.hg.1 ENST00000414823.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600013564.hg.1 ENST00000440335.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008203.hg.1 ENST00000442381.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600165070.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600165070.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600165070.hg.1 NM_001016.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600165070.hg.1 TC0600009488.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600165070.hg.1 ENST00000230050.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600165070.hg.1 ENST00000484616.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600165070.hg.1 NM_001016.3 Refseq
          Affymetrix Human Clariom D Assay TC0500011293.hg.1 ENST00000510537.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400010828.hg.1 ENST00000461060.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500008301.hg.1 ENST00000492017.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000011207.hg.1 ENST00000439728.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011627.hg.1 ENST00000428553.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011627.hg.1 ENST00000428553.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700009523.hg.1 ENST00000466297.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008188.hg.1 ENST00000465542.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300008275.hg.1 ENST00000419579.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011609.hg.1 ENST00000497190.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900006638.hg.1 ENST00000437701.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0