Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04185568_m1
          See other DDTL GE Assays ›
          Gene Symbol:
          DDTL
          Gene Name:
          D-dopachrome tautomerase-like
          Gene Aliases:
          KB-226F1.2
          Chromosome Location:
          Chr.22: 23966221 - 23972559 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04185568_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DDTL
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001084393.1 NP_001077862.1 2 - 3 360 85
          GenBank mRNA AK296110.1 - 2 - 3 310 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2200006874.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC22000141.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Process(es)

          lyase activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11444676 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32122663 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11444670 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32122672 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32122668 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11444666 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11444668 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17267709 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32122662 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32122671 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 53320 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 70949 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 34912 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 11984 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 717379 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 761337 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 934360 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 195847 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 401050 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 269327 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 303228 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3340205 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1526 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1527 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 59168 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P21291595 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P03605949 - -
          Agilent Human Genome CGH + SNP Microarray A_14_P115891 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3340205 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3340205 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32122662 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444666 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32122663 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444676 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32122672 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17267709 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444668 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444676 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444666 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32122663 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17267709 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32122672 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32122662 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444668 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P115891 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P112167 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P138267 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 130387 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 62263 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 35205 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 82283 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P112167 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P115891 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21291596 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11444666 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11444667 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11444673 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11444680 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11444683 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11444670 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11444677 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11444668 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P115891 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P112167 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P21291596 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8071808 - -
          Affymetrix Human Gene 1.0 ST Array 8074970 - -
          Affymetrix Human Gene 1.0 ST Array 8074971 - -
          Affymetrix Human Gene 1.0 ST Array 8071807 - -
          Affymetrix Human Gene 1.1 ST Array 8071806 - -
          Affymetrix Human Gene 1.1 ST Array 8074969 - -
          Affymetrix Human Genome U219 Array 11760187_PM_at - -
          Affymetrix Human Genome U219 Array 11740150_PM_at - -
          Affymetrix Human Genome U219 Array 11740150_at - -
          Affymetrix Human Genome U219 Array 11760188_PM_x_at - -
          Affymetrix Human Genome U219 Array 11760187_at - -
          Affymetrix Human Genome U219 Array 11761623_PM_at - -
          Affymetrix Human Genome U219 Array 11761623_at - -
          Affymetrix Human Genome U219 Array 11760188_x_at - -
          Affymetrix Human Transcriptome Array 2.0 TC2200006874.hg.1 ENST00000215770.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006874.hg.1 ENST00000215770.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006874.hg.1 ENST00000215770.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006874.hg.1 ENST00000215770.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006874.hg.1 NM_001084393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200006874.hg.1 NM_001084393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200006874.hg.1 NM_001084393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200006874.hg.1 NM_001084393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200006874.hg.1 NM_001084393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200006874.hg.1 NM_001084393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200006874.hg.1 NM_001144931.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200006874.hg.1 NM_001144931.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200006874.hg.1 NM_001355.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200006874.hg.1 NM_001355.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200006874.hg.1 NM_001355.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22002573.hg.1 TC22000141.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22002573.hg.1 NM_001084393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22002574.hg.1 TC22000141.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22002574.hg.1 ENST00000215770.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22002574.hg.1 NM_001084393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22002577.hg.1 TC22000141.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22002577.hg.1 ENST00000215770.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22002577.hg.1 NM_001084393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22002575.hg.1 TC22000141.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22002575.hg.1 ENST00000215770.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22002575.hg.1 NM_001084393.1 Refseq
          Affymetrix Human Clariom D Assay TC2200006874.hg.1 ENST00000215770.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200006874.hg.1 ENST00000215770.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200006874.hg.1 ENST00000215770.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200006874.hg.1 ENST00000215770.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200006874.hg.1 NM_001084393.1 Refseq
          Affymetrix Human Clariom D Assay TC2200006874.hg.1 NM_001084393.1 Refseq
          Affymetrix Human Clariom D Assay TC2200006874.hg.1 NM_001084393.1 Refseq
          Affymetrix Human Clariom D Assay TC2200006874.hg.1 NM_001084393.1 Refseq
          Affymetrix Human Clariom D Assay TC2200006874.hg.1 NM_001084393.1 Refseq
          Affymetrix Human Clariom D Assay TC2200006874.hg.1 NM_001084393.1 Refseq
          Affymetrix Human Clariom D Assay TC2200006874.hg.1 NM_001144931.2 Refseq
          Affymetrix Human Clariom D Assay TC2200006874.hg.1 NM_001144931.2 Refseq
          Affymetrix Human Clariom D Assay TC2200006874.hg.1 NM_001355.3 Refseq
          Affymetrix Human Clariom D Assay TC2200006874.hg.1 NM_001355.3 Refseq
          Affymetrix Human Clariom D Assay TC2200006874.hg.1 NM_001355.3 Refseq
          Affymetrix Human Clariom D Assay PSR2200147476.hg.1 ENST00000215770.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200147476.hg.1 NM_001084393.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200147476.hg.1 TC2200006874.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200147476.hg.1 NM_001084393.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200147476.hg.1 ENST00000215770.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200147467.hg.1 NM_001084393.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200147467.hg.1 TC2200006874.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200147467.hg.1 NM_001084393.1 Refseq
          Human Genome U95 Set 374_f_at - -
          Human Genome U95 Set 374_PM_f_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A020217 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A020217 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3244439 CR606018.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3244439 NM_001084393.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3244439 AI686044.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3244439 CR606018.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3244439 NM_001084393.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3244439 AI686044.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3244439 CR606018.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3244439 NM_001084393.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3244439 AI686044.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3244439 CR606018.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3244439 NM_001084393.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3244439 AI686044.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0