Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003043.hg.1 |
ENST00000312423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003043.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003043.hg.1 |
TC19000197.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003048.hg.1 |
ENST00000312423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003048.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003048.hg.1 |
TC19000197.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007026.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007026.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007026.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007026.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007026.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007026.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007026.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007026.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007026.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007026.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007026.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007026.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003044.hg.1 |
ENST00000312423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003044.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003044.hg.1 |
TC19000197.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003047.hg.1 |
ENST00000312423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003047.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003047.hg.1 |
TC19000197.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008808.hg.1 |
ENST00000583623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150374.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150374.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900150374.hg.1 |
TC1900007026.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900150374.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900150374.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150377.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150377.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900150377.hg.1 |
TC1900007026.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900150377.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150377.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007026.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007026.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007026.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007026.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007026.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007026.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007026.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007026.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007026.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007026.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007026.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007026.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008808.hg.1 |
ENST00000583623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150375.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150375.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900150375.hg.1 |
TC1900007026.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900150375.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150375.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900150378.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150378.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900150378.hg.1 |
TC1900007026.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900150378.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150378.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900150376.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150376.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900150376.hg.1 |
TC1900007026.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900150376.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150376.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900150379.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150379.hg.1 |
NM_175871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900150379.hg.1 |
TC1900007026.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900150379.hg.1 |
ENST00000312423.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150379.hg.1 |
NM_175871.3 |
Refseq |