Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04231446_s1
          See other MIR106A GE Assays ›
          Gene Symbol:
          MIR106A
          Gene Name:
          microRNA 106a
          Gene Aliases:
          MIRN106A, mir-106, mir-106a
          Chromosome Location:
          Chr.X: 134170198 - 134170278 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04231446_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIR106A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029523.1 1 34 81
          GenBank mRNA AF480549.1 - 1 34 81

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 18b MIR18B MIRN18B,
          hsa-mir-18b,
          mir-18b
          -87.0 Sense Chr. X - 134170041 - 134170111 on NCBI Build GRCh38
          Downstream coiled-coil domain containing 160 CCDC160 66.8kb Anti-sense Chr. X - 134237047 - 134246207 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
          This assay targets the same location as Clariom D transcript cluster(s) TC0X00010862.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X001386.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967896 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967919 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699781 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967902 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967926 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699737 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967925 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699788 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699718 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967914 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699766 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699711 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699753 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967908 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967918 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699790 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699712 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699760 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967901 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699759 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699756 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967916 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967910 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699751 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699765 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699764 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699717 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699749 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699770 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967911 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699768 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699778 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699776 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699777 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699780 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699783 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967882 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699773 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699786 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699782 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699785 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699774 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967905 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699757 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699761 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699716 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967886 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699758 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967888 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699714 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967903 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967923 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699771 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699762 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699784 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699719 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699772 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967894 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967898 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699776 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699762 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699719 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699777 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967888 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699788 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967901 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699781 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699790 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699780 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967911 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699749 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699712 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699766 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967919 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699758 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967908 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699772 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967886 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967923 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967903 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699714 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699757 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967882 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967910 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967905 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699778 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699785 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699756 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699718 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699765 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967925 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699773 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699759 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699782 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699761 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967918 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699774 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967896 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699760 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699753 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967898 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699770 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699764 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699786 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967902 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699783 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699768 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967914 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699751 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967894 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699716 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967926 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699737 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699717 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11967916 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699784 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699711 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32699771 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8175253 - -
          Affymetrix Human Gene 1.1 ST Array 8175252 - -
          Affymetrix Human Transcriptome Array 2.0 TC0X00010862.hg.1 ENST00000384870.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010862.hg.1 NR_029523.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X018530.hg.1 ENST00000384870.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X018530.hg.1 NR_029523.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X018530.hg.1 TC0X001386.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0X00173759.hg.1 ENST00000384870.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00173759.hg.1 NR_029523.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00173759.hg.1 TC0X00010862.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00173759.hg.1 NR_029523.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00173759.hg.1 ENST00000384870.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010862.hg.1 ENST00000384870.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010862.hg.1 NR_029523.1 Refseq
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3309409 NR_029523.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3309409 AF480549.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3309409 NR_029523.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3309409 AF480549.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3309409 NR_029523.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3309409 AF480549.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3309409 NR_029523.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3309409 AF480549.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0