Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04231455_s1
          See other MIR147A GE Assays ›
          Gene Symbol:
          MIR147A
          Gene Name:
          microRNA 147a
          Gene Aliases:
          MIR147, MIRN147, hsa-mir-147a
          Chromosome Location:
          Chr.9: 120244979 - 120245050 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04231455_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIR147A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029604.1 1 37 69
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CD675758.1 - 1 292 Chr. 9 - 120244966 - 120245303 [-] on Build GRCh38 69

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream BMP/retinoic acid inducible neural specific 1 BRINP1 DBC1,
          DBCCR1,
          FAM5A
          -875.5kb Sense Chr. 9 - 119166630 - 119369461 on NCBI Build GRCh38
          Downstream CDK5 regulatory subunit associated protein 2 CDK5RAP2 C48,
          Cep215,
          MCPH3
          143.8kb Sense Chr. 9 - 120388869 - 120580170 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
          This assay targets the same location as Clariom D transcript cluster(s) TC0900011375.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09001534.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282505 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937727 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937716 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282523 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937733 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937744 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16259254 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937736 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282512 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282493 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282529 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937711 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937714 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937715 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282524 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937737 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282491 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937742 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937745 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282496 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282506 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937719 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937749 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282482 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937726 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937746 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937731 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937709 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16259253 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937710 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937713 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937740 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282530 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937721 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282499 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937725 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937732 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937749 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937727 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282512 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937709 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937742 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937736 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937711 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282524 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282529 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16259253 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282505 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937714 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937740 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937715 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282491 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937731 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282506 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937721 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937725 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282496 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937732 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282493 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16259254 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937719 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937745 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937746 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937744 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937710 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937726 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937716 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937713 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282530 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282523 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282482 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937733 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27282499 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06937737 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.1 ST Array 8163729 - -
          Affymetrix Human Transcriptome Array 2.0 TC0900011375.hg.1 ENST00000385079.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011375.hg.1 NR_029604.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09019147.hg.1 ENST00000385079.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09019147.hg.1 NR_029604.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09019147.hg.1 TC09001534.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0900177428.hg.1 ENST00000385079.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177428.hg.1 NR_029604.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900177428.hg.1 TC0900011375.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900177428.hg.1 NR_029604.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900177428.hg.1 ENST00000385079.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011375.hg.1 ENST00000385079.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011375.hg.1 NR_029604.1 Refseq
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3309389 NR_029604.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3309389 NR_029604.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3309389 NR_029604.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3309389 NR_029604.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0