Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04231535_s1
          See other MIR128-2 GE Assays ›
          Gene Symbol:
          MIR128-2
          Gene Name:
          microRNA 128-2
          Gene Aliases:
          MIR128B, MIRN128-2, MIRN128B, mir-128-2, mir-128b
          Chromosome Location:
          Chr.3: 35744476 - 35744559 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04231535_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIR128-2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029824.1 1 43 70

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC101928135 LOC101928135 -350.5kb Anti-sense Chr. 3 - 34875795 - 35394023 on NCBI Build GRCh38
          Downstream SH3 and cysteine rich domain STAC STAC1 635.9kb Sense Chr. 3 - 36380455 - 36548007 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
          This assay targets the same location as Clariom D transcript cluster(s) TC0300007017.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03000172.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301687 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167769 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301684 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167770 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301646 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167771 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301630 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301692 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167747 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301691 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301631 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167774 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301638 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15427143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301686 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301650 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301664 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301679 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167754 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167755 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167753 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301656 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167762 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167781 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301653 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301667 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167759 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167812 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301633 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301675 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301666 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301625 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167766 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301642 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301670 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301661 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301669 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167784 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301673 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301674 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301668 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167779 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301681 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301657 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301626 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301665 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301677 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301639 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167771 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301661 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301673 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167781 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301666 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167755 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167762 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167754 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301630 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167769 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301631 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301653 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301677 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301657 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301638 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301625 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15427143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301656 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301675 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301674 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167784 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301626 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167770 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301646 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301686 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301642 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167779 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301670 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301691 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167759 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301639 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301684 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301665 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301679 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301633 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167812 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167753 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301692 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167766 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167774 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02167747 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301667 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301650 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301668 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301687 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301664 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301669 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22301681 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.1 ST Array 8078527 - -
          Affymetrix Human Transcriptome Array 2.0 PSR03003230.hg.1 ENST00000384893.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03003230.hg.1 NR_029824.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03003230.hg.1 TC03000172.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0300007017.hg.1 ENST00000384893.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007017.hg.1 NR_029824.1 Refseq
          Affymetrix Human Clariom D Assay TC0300007017.hg.1 ENST00000384893.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007017.hg.1 NR_029824.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300148943.hg.1 ENST00000384893.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300148943.hg.1 NR_029824.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300148943.hg.1 TC0300007017.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300148943.hg.1 ENST00000384893.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300148943.hg.1 NR_029824.1 Refseq
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3308911 NR_029824.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3308911 NR_029824.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308911 NR_029824.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308911 NR_029824.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0