Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04231563_s1
          See other MIR337 GE Assays ›
          Gene Symbol:
          MIR337
          Gene Name:
          microRNA 337
          Gene Aliases:
          MIRN337, hsa-mir-337, mir-337
          Chromosome Location:
          Chr.14: 100874493 - 100874585 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04231563_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIR337
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029889.1 1 49 63

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 493 MIR493 MIRN493,
          hsa-mir-493,
          mir-493
          -5.3kb Sense Chr. 14 - 100869060 - 100869148 on NCBI Build GRCh38
          Downstream microRNA 665 MIR665 MIRN665,
          hsa-mir-665,
          mir-665
          448.0 Sense Chr. 14 - 100875033 - 100875104 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
          This assay targets the same location as Clariom D transcript cluster(s) TC1400008235.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14000652.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979004 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516695 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516714 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979006 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516700 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516698 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516708 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516715 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516717 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979028 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516693 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979030 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979024 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979009 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516705 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979018 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516716 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516719 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516691 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516700 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516698 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979028 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516691 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516717 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979009 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516708 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516716 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516693 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516719 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516714 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516715 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979004 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516705 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979018 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979024 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09516695 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979030 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979006 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1400008235.hg.1 ENST00000362281.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008235.hg.1 NR_029889.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14007979.hg.1 ENST00000362281.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14007979.hg.1 NR_029889.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14007979.hg.1 TC14000652.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1400158708.hg.1 ENST00000362281.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1400158708.hg.1 NR_029889.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400158708.hg.1 TC1400008235.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400158708.hg.1 NR_029889.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400158708.hg.1 ENST00000362281.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008235.hg.1 ENST00000362281.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008235.hg.1 NR_029889.1 Refseq
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3308415 NR_029889.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3308415 NR_029889.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308415 NR_029889.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308415 NR_029889.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0