Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04231583_s1
          See other MIR448 GE Assays ›
          Gene Symbol:
          MIR448
          Gene Name:
          microRNA 448
          Gene Aliases:
          MIRN448, hsa-mir-448, miRNA448
          Chromosome Location:
          Chr.X: 114823454 - 114823564 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04231583_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIR448
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029955.1 1 58 65

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 1911 MIR1911 MIRN1911,
          hsa-mir-1911,
          mir-1911
          -60.2kb Sense Chr. X - 114763184 - 114763263 on NCBI Build GRCh38
          Downstream interleukin 13 receptor subunit alpha 2 IL13RA2 CD213A2,
          CT19,
          IL-13R,
          IL13BP
          180.4kb Anti-sense Chr. X - 115003975 - 115017644 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
          This assay targets the same location as Clariom D transcript cluster(s) TC0X00008188.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X000563.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17388905 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630032 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11899995 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630041 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630006 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630018 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630050 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630040 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630033 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630025 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11900016 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630030 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630031 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11900007 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11900015 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630013 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630051 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630014 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11899997 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630046 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17388919 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630019 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17388904 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11900032 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630023 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11900005 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17388912 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630039 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630021 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630016 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630022 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630010 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630044 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11900030 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630048 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630015 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630045 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17388906 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630008 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630038 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630039 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11900016 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11900032 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630023 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17388912 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630046 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630048 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630022 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630015 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630025 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630031 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630008 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630044 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630051 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11900007 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11900005 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630006 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11899997 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630019 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17388904 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630014 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630021 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17388905 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630033 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11900015 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17388906 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630040 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17388919 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630010 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630016 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630030 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11899995 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630018 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11900030 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630045 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630032 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630013 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630041 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630038 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32630050 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR0X007435.hg.1 ENST00000362131.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X007435.hg.1 NR_029955.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X007435.hg.1 TC0X000563.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0X00008188.hg.1 ENST00000362131.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008188.hg.1 NR_029955.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00155843.hg.1 ENST00000362131.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00155843.hg.1 NR_029955.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00155843.hg.1 TC0X00008188.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00155843.hg.1 NR_029955.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00155843.hg.1 ENST00000362131.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008188.hg.1 ENST00000362131.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008188.hg.1 NR_029955.1 Refseq
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3309844 NR_029955.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3309844 NR_029955.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0