Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04231647_s1
          See other MIR499A GE Assays ›
          Gene Symbol:
          MIR499A MIR499B
          Gene Name:
          microRNA 499a
          microRNA 499b
          Gene Aliases:
          MIR499, MIRN499, hsa-mir-499a, mir-499a
          MIR499A, hsa-mir-499b
          Chromosome Location:
          Chr.20: 34990376 - 34990497 on Build GRCh38
          Chromosome Location:
          Chr.20: 34990400 - 34990472 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs04231647_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIR499A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_030223.1 1 59 70
          Gene Symbol: MIR499B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_039912.1 1 70
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AK098707.1 - 1 Chr. 20 - 34988975 - 34990998 [-] on Build GRCh38 70

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream myosin heavy chain 7B MYH7B MHC14,
          MYH14
          Sense Chr. 20 - 34955835 - 35002437 on NCBI Build GRCh38
          Downstream transient receptor potential cation channel subfamily C member 4 associated protein TRPC4AP C20orf188,
          PPP1R158,
          TRRP4AP,
          TRUSS
          11.9kb Anti-sense Chr. 20 - 35002404 - 35092837 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
          This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
          This assay targets the same location as Clariom D transcript cluster(s) TC2000007207.hg.1, TC2000008954.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000247.hg.1, TC20000790.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143381 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143341 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791090 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143345 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143350 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143355 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791059 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791043 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143373 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143337 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143340 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791084 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143329 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791035 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17188812 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143324 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791040 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791042 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143378 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791072 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143349 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17188793 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791085 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143376 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143359 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143363 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143360 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143361 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143357 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143348 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143342 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143352 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791079 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143344 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143353 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791049 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791047 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791088 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17188797 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791056 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143328 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791036 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791087 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143335 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791034 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791095 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17188797 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791043 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791072 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143350 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143328 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143344 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143329 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791088 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143341 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143324 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791056 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791035 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143381 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143345 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791049 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791079 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791084 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143355 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17188793 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791034 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791095 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143352 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143373 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791087 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791047 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143342 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143348 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143360 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143335 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143340 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143361 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143337 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791085 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17188812 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143357 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143353 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791090 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791042 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143376 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143349 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791059 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143359 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791040 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143378 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31791036 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11143363 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR20003271.hg.1 TC20000247.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20003271.hg.1 ENST00000384903.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20003271.hg.1 NR_030223.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007207.hg.1 ENST00000384903.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007207.hg.1 NR_030223.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20010552.hg.1 TC20000790.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20010552.hg.1 NR_039912.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007207.hg.1 ENST00000384903.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007207.hg.1 NR_030223.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000162340.hg.1 NR_039912.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000162340.hg.1 TC2000008954.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000162340.hg.1 NR_039912.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000149553.hg.1 ENST00000384903.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000149553.hg.1 NR_030223.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000149553.hg.1 TC2000007207.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000149553.hg.1 NR_030223.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000149553.hg.1 ENST00000384903.1 Ensembl
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3309619 NR_030223.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3309619 NR_030223.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3309619 NR_030223.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3309619 NR_030223.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0