Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04231687_s1
          See other MIR589 GE Assays ›
          Gene Symbol:
          MIR589
          Gene Name:
          microRNA 589
          Gene Aliases:
          MIRN589, hsa-mir-589, mir-589
          Chromosome Location:
          Chr.7: 5495819 - 5495917 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04231687_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIR589
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_030318.1 1 46 69
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CD652196.1 - 1 329 Chr. 7 - 5495389 - 5496199 [-] on Build GRCh38 69

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC100129484 LOC100129484 -64.8kb Sense Chr. 7 - 5426281 - 5429053 on NCBI Build GRCh38
          Downstream uncharacterized LOC221946 LOC221946 17.9kb Anti-sense Chr. 7 - 5513854 - 5525545 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
          This assay targets the same location as Clariom D transcript cluster(s) TC0700010165.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07001119.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15905360 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15905362 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278250 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25493544 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278258 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278267 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278253 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25493521 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278252 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25493531 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278249 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25493526 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278273 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278266 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278260 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278254 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278270 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278262 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278256 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278264 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278251 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15905354 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15905361 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15905357 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278263 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278259 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278247 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25493521 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278262 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278253 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278266 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278252 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278270 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278260 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278273 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278267 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278247 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278264 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278256 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15905360 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15905354 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25493526 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278259 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278250 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15905361 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278254 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15905357 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278258 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278251 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278263 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25493544 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25493531 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05278249 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15905362 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0700010165.hg.1 ENST00000385238.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010165.hg.1 NR_030318.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07017233.hg.1 ENST00000385238.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07017233.hg.1 NR_030318.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07017233.hg.1 TC07001119.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0700172202.hg.1 ENST00000385238.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700172202.hg.1 NR_030318.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700172202.hg.1 TC0700010165.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700172202.hg.1 NR_030318.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700172202.hg.1 ENST00000385238.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010165.hg.1 ENST00000385238.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010165.hg.1 NR_030318.1 Refseq
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3308083 NR_030318.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3308083 NR_030318.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308083 NR_030318.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308083 NR_030318.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0