Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02025535.hg.1 |
NR_036226.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02025535.hg.1 |
TC02001585.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011764.hg.1 |
ENST00000578394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011764.hg.1 |
NR_036226.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011764.hg.1 |
ENST00000578394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011764.hg.1 |
NR_036226.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200185503.hg.1 |
ENST00000578394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200185503.hg.1 |
NR_036226.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200185503.hg.1 |
TC0200011764.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200185503.hg.1 |
NR_036226.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200185503.hg.1 |
ENST00000578394.1 |
Ensembl |