Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04232004_s1
          See other LOC100499489 GE Assays ›
          Gene Symbol:
          LOC100499489
          Gene Name:
          uncharacterized LOC100499489
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 22435425 - 22437929 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04232004_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100499489
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_036533.1 1 2049 103
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AA417966.1 - 1 Chr. 10 - 22435433 - 22435936 [+] on Build GRCh38 103
          GenBankmRNA CR740398.1 - 1 342 Chr. 10 - 22435691 - 22436222 [-] on Build GRCh38 103
          GenBankmRNA AA470114.1 - 1 342 Chr. 10 - 22435756 - 22436223 [-] on Build GRCh38 103
          GenBankmRNA W93459.1 - 136 Chr. - - [-] on Build 103
          GenBankmRNA CK005617.1 - 1 315 Chr. 10 - 22435522 - 22436193 [-] on Build GRCh38 103
          GenBankmRNA BM712770.1 - 1 86 Chr. 10 - 22435519 - 22435966 [-] on Build GRCh38 103
          GenBankmRNA CK902567.1 - 1 Chr. 10 - 22435430 - 22436006 [+] on Build GRCh38 103
          GenBankmRNA BX115098.1 - 1 Chr. 10 - 22435430 - 22436165 [+] on Build GRCh38 103
          GenBankmRNA DB329684.1 - 1 Chr. 10 - 22435433 - 22435993 [+] on Build GRCh38 103
          GenBankmRNA AI916303.1 - 1 Chr. 10 - 22435433 - 22436038 [+] on Build GRCh38 103
          GenBankmRNA BF197159.1 - 1 Chr. 10 - 22435432 - 22435967 [+] on Build GRCh38 103

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream sperm associated antigen 6 SPAG6 CT141,
          Repro-SA-1,
          pf16
          -17.8kb Anti-sense Chr. 10 - 22345416 - 22417610 on NCBI Build GRCh38
          Downstream phosphatidylinositol-5-phosphate 4-kinase type 2 alpha PIP4K2A PI5P4KA,
          PIP5K2A,
          PIP5KII-alpha,
          PIP5KIIA,
          PIPK
          96.9kb Sense Chr. 10 - 22534837 - 22714574 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000010020.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10001100.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10013824.hg.1 NR_036533.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10013824.hg.1 TC10001100.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1000168353.hg.1 NR_036533.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000168353.hg.1 TC1000010020.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000168353.hg.1 NR_036533.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000168352.hg.1 NR_036533.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000168352.hg.1 TC1000010020.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000168352.hg.1 NR_036533.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000168348.hg.1 NR_036533.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000168348.hg.1 TC1000010020.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000168348.hg.1 NR_036533.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000168350.hg.1 NR_036533.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000168350.hg.1 TC1000010020.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000168350.hg.1 NR_036533.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000168351.hg.1 NR_036533.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000168351.hg.1 TC1000010020.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000168351.hg.1 NR_036533.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000168349.hg.1 NR_036533.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000168349.hg.1 TC1000010020.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000168349.hg.1 NR_036533.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fcd8d4bc9-cwdc5:80/100.66.130.62:80.
          git-commit: a06ae585138c3108a562191b4d7dc526e77a8a21
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0