Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04232307_m1
          See other LOC285626 GE Assays ›
          Gene Symbol:
          LOC285626
          Gene Name:
          uncharacterized LOC285626
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 159331518 - 159362834 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04232307_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC285626
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_037889.1 2 - 3 743 94
          GenBank mRNA AK097548.1 - 2 - 3 742 94
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DA733091.1 - 2 - 3 411 Chr. 5 - 159331939 - 159416624 [+] on Build GRCh38 94

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream interleukin 12B IL12B CLMF,
          CLMF2,
          IL-12B,
          IMD28,
          IMD29,
          NKSF,
          NKSF2
          -2.3kb Anti-sense Chr. 5 - 159314783 - 159330473 on NCBI Build GRCh38
          Downstream uncharacterized LOC285627 LOC285627 101.0kb Anti-sense Chr. 5 - 159448556 - 159466276 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500009253.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05000887.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406917 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586458 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406895 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406956 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406935 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586453 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406893 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406957 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586404 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586451 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586454 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586457 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586445 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406891 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406926 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586424 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586447 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586396 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406900 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406958 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586409 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406884 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406894 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586399 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586436 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406939 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586466 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406916 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586440 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586435 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586438 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406950 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406949 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406909 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406904 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586443 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406951 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406902 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406877 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406915 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586455 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406929 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406881 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406914 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406907 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586402 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406933 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406947 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586413 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586401 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406906 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586425 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24586414 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04406961 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 129344 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 142107 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 39915 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 614756 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 462375 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 177264 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 410582 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 233730 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 277719 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 727218 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 805554 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 911236 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 499428 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 289252 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 397988 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3525062 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 24746 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P01377066 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P37434421 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P17368078 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P15463902 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3525062 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3525062 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P17368078 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P37434421 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P17368064 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P17368078 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P01377084 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 73649 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 125476 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 21928 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17368070 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01377066 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P24648207 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01377086 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17368111 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01377082 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17368074 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01377084 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P37434421 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17368064 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17368061 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17368078 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P17368078 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P37434421 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P01377066 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P01377086 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P01377084 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P17368064 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR05012819.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05012819.hg.1 NR_037889.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05012819.hg.1 TC05000887.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05012821.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05012821.hg.1 NR_037889.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05012821.hg.1 TC05000887.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 NR_037889.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 NR_037889.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 NR_037889.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 NR_037889.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 NR_037889.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 NR_037889.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009253.hg.1 NR_037889.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05012820.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05012820.hg.1 NR_037889.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05012820.hg.1 TC05000887.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 NR_037889.1 Refseq
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 NR_037889.1 Refseq
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 NR_037889.1 Refseq
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 NR_037889.1 Refseq
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 NR_037889.1 Refseq
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 NR_037889.1 Refseq
          Affymetrix Human Clariom D Assay TC0500009253.hg.1 NR_037889.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500164736.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500164736.hg.1 NR_037889.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500164736.hg.1 TC0500009253.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500164736.hg.1 NR_037889.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500164736.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500164737.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500164737.hg.1 NR_037889.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500164737.hg.1 TC0500009253.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500164737.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500164737.hg.1 NR_037889.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500164738.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500164738.hg.1 NR_037889.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500164738.hg.1 TC0500009253.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500164738.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500164738.hg.1 NR_037889.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500164735.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500164735.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500164735.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500164735.hg.1 NR_037889.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500164735.hg.1 TC0500009253.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500164735.hg.1 ENST00000515337.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500164735.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500164735.hg.1 ENST00000521472.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500164735.hg.1 NR_037889.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A029096 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A029096 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fcd8d4bc9-cwdc5:80/100.66.130.62:80.
          git-commit: a06ae585138c3108a562191b4d7dc526e77a8a21
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0