Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT HG-U133+ PM Array Plate |
230277_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008956.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008956.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008956.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008956.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008956.hg.1 |
TC07003290.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000429679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000429679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000429679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000429679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000609449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000609449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000609449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000609449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000609449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000609449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
ENST00000609449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013424.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008957.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008957.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008957.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008957.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008957.hg.1 |
TC07003290.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0700198156.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198156.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198156.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198156.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198156.hg.1 |
ENST00000609449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198156.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198156.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198156.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198156.hg.1 |
TC0700013424.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700198156.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198156.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198156.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198156.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198156.hg.1 |
ENST00000609449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198156.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198156.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198156.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198158.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198158.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198158.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198158.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198158.hg.1 |
ENST00000609449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198158.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198158.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198158.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198158.hg.1 |
TC0700013424.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700198158.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198158.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198158.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198158.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198158.hg.1 |
ENST00000609449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198158.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198158.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198158.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198159.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198159.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198159.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198159.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198159.hg.1 |
ENST00000609449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198159.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198159.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198159.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198159.hg.1 |
TC0700013424.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700198159.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198159.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198159.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198159.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198159.hg.1 |
ENST00000609449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198159.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198159.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198159.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198160.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198160.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198160.hg.1 |
ENST00000609449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198160.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198160.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198160.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198160.hg.1 |
TC0700013424.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700198160.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198160.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198160.hg.1 |
ENST00000609449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198160.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198160.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198160.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000429679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000429679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000429679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000429679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000431679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000453227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000609449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000609449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000609449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000609449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000609449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000609449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
ENST00000609449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195542.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013424.hg.1 |
NM_001195543.2 |
Refseq |
Human Genome U95 Set |
72261_s_at |
- |
- |
Human Genome U95 Set |
72261_PM_s_at |
- |
- |
Human Genome U95 Set |
77666_PM_at |
- |
- |
Human Genome U95 Set |
77666_at |
- |
- |
Human Genome U133 Set |
230277_PM_at |
- |
- |
Human Genome U133 Set |
230277_at |
- |
- |