Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002738.hg.1 |
ENST00000219022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002738.hg.1 |
ENST00000489054.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002738.hg.1 |
NM_006418.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002738.hg.1 |
TC13000228.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007256.hg.1 |
ENST00000219022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007256.hg.1 |
ENST00000219022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007256.hg.1 |
ENST00000219022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007256.hg.1 |
ENST00000219022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007256.hg.1 |
ENST00000219022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007256.hg.1 |
ENST00000219022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007256.hg.1 |
ENST00000219022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007256.hg.1 |
ENST00000219022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007256.hg.1 |
NM_006418.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007256.hg.1 |
NM_006418.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007256.hg.1 |
NM_006418.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007256.hg.1 |
NM_006418.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007256.hg.1 |
NM_006418.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007256.hg.1 |
NM_006418.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007256.hg.1 |
NM_006418.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007256.hg.1 |
NM_006418.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149308.hg.1 |
ENST00000219022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149308.hg.1 |
NM_006418.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149308.hg.1 |
TC1300007256.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300149308.hg.1 |
NM_006418.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149308.hg.1 |
ENST00000219022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007256.hg.1 |
ENST00000219022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007256.hg.1 |
ENST00000219022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007256.hg.1 |
ENST00000219022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007256.hg.1 |
ENST00000219022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007256.hg.1 |
ENST00000219022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007256.hg.1 |
ENST00000219022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007256.hg.1 |
ENST00000219022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007256.hg.1 |
ENST00000219022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007256.hg.1 |
NM_006418.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007256.hg.1 |
NM_006418.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007256.hg.1 |
NM_006418.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007256.hg.1 |
NM_006418.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007256.hg.1 |
NM_006418.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007256.hg.1 |
NM_006418.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007256.hg.1 |
NM_006418.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007256.hg.1 |
NM_006418.4 |
Refseq |