Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs04261500_m1
See other IKZF1 GE Assays ›
Gene Symbol:
IKZF1
Gene Name:
IKAROS family zinc finger 1
Gene Aliases:
CVID13, Hs.54452, IK1, IKAROS, LYF1, LyF-1, PPP1R92, PRO0758, ZNFN1A1
Chromosome Location:
Chr.7: 50303453 - 50405101 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs04261500_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: IKZF1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001220765.2
NP_001207694.1
2 - 3
309
74
NM_001220767.2
NP_001207696.1
1 - 2
55
74
NM_001220768.2
NP_001207697.1
1 - 2
55
74
NM_001220770.2
NP_001207699.1
1 - 2
55
74
NM_001220771.2
NP_001207700.1
1 - 2
55
74
NM_001291837.1
NP_001278766.1
2 - 3
282
74
NM_001291838.1
NP_001278767.1
2 - 3
282
74
NM_001291839.1
NP_001278768.1
2 - 3
87
74
NM_001291840.1
NP_001278769.1
1 - 2
55
74
NM_001291841.1
NP_001278770.1
1 - 2
55
74
NM_001291842.1
NP_001278771.1
1 - 2
55
74
NM_001291843.1
NP_001278772.1
1 - 2
55
74
NM_001291844.1
NP_001278773.1
1 - 2
55
74
NM_001291845.1
NP_001278774.1
2 - 3
309
74
NM_001291846.1
NP_001278775.1
2 - 3
309
74
NM_001291847.1
NP_001278776.1
2 - 3
282
74
NM_006060.5
NP_006051.1
2 - 3
309
74
XM_011515058.2
XP_011513360.1
2 - 3
139
74
XM_011515059.2
XP_011513361.1
2 - 3
141
74
XM_011515060.2
XP_011513362.1
2 - 3
3289
74
XM_011515061.2
XP_011513363.1
2 - 3
137
74
XM_011515062.2
XP_011513364.1
2 - 3
419
74
XM_011515063.2
XP_011513365.1
2 - 3
336
74
XM_011515064.2
XP_011513366.1
2 - 3
3967
74
XM_011515065.2
XP_011513367.1
2 - 3
104
74
XM_011515066.2
XP_011513368.1
2 - 3
208
74
XM_011515067.2
XP_011513369.1
2 - 3
141
74
XM_011515068.2
XP_011513370.1
2 - 3
336
74
XM_011515069.2
XP_011513371.1
2 - 3
3964
74
XM_011515070.2
XP_011513372.1
2 - 3
140
74
XM_011515071.2
XP_011513373.1
2 - 3
139
74
XM_011515072.2
XP_011513374.1
2 - 3
137
74
XM_011515073.2
XP_011513375.1
2 - 3
419
74
XM_011515074.2
XP_011513376.1
2 - 3
140
74
XM_011515075.2
XP_011513377.1
2 - 3
419
74
XM_011515076.2
XP_011513378.1
2 - 3
363
74
XM_011515077.2
XP_011513379.1
2 - 3
140
74
XM_011515078.2
XP_011513380.1
2 - 3
143
74
XM_017011667.1
XP_016867156.1
2 - 3
336
74
XM_017011668.1
XP_016867157.1
2 - 3
103
74
XM_017011669.1
XP_016867158.1
2 - 3
104
74
XM_017011670.1
XP_016867159.1
2 - 3
465
74
GenBank mRNA
AK303586.1
-
2 - 3
241
74
AY377974.1
-
2 - 3
209
74
BC018349.1
-
2 - 3
233
4338834
74
BM148203.1
-
2 - 3
301
74
BT009836.1
-
1 - 2
41
74
BU537824.1
-
2 - 3
217
74
DB122945.1
-
2 - 3
282
74
DB148957.1
-
2 - 3
309
74
DC391353.1
-
2 - 3
93
74
JX459579.1
-
2 - 3
209
74
U40462.1
-
2 - 3
209
74
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length between 71 and 85
Mammalian Gene Collection
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene IKZF1
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription by RNA polymerase II
transcription, DNA-templated
cell cycle
mesoderm development
lymphocyte differentiation
erythrocyte differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
transcription cis-regulatory region binding
DNA binding
DNA-binding transcription factor activity
protein binding
metal ion binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0