Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04273253_s1
          See other MIR193A GE Assays ›
          Gene Symbol:
          MIR193A
          Gene Name:
          microRNA 193a
          Gene Aliases:
          MIRN193, MIRN193A, mir-193a
          Chromosome Location:
          Chr.17: 31559996 - 31560083 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04273253_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIR193A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029710.1 1 44 73

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RAB11 family interacting protein 4 RAB11FIP4 RAB11-FIP4 -21.8kb Sense Chr. 17 - 31391624 - 31538217 on NCBI Build GRCh38
          Downstream microRNA 4725 MIR4725 mir-4725 15.2kb Sense Chr. 17 - 31575269 - 31575358 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
          This assay targets the same location as Clariom D transcript cluster(s) TC1700007485.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17000365.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850043 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10283998 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284001 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850028 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284006 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850035 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850027 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284004 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284008 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284023 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850016 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16969427 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284012 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284026 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16969431 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850052 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850032 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850069 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284013 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284042 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284009 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284021 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850031 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10283989 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284007 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10283992 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850018 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284020 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850057 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850054 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850049 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284002 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850021 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284036 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284028 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10283996 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284040 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284038 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284047 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284046 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284044 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284027 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284032 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850041 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10283994 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10283998 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850052 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850043 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16969431 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850016 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850049 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284038 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284009 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850027 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284023 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284002 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284036 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284027 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284042 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850035 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850069 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284007 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284001 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10283994 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850032 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850018 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284040 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284012 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284047 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10283992 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284044 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284021 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284008 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10283989 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284028 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850021 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284020 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284046 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284004 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284013 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10283996 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850041 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850054 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850028 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284026 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850057 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16969427 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284032 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10284006 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30850031 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.1 ST Array 8006321 - -
          Affymetrix Human Transcriptome Array 2.0 TC1700007485.hg.1 ENST00000384882.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007485.hg.1 NR_029710.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17004631.hg.1 ENST00000384882.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17004631.hg.1 NR_029710.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17004631.hg.1 TC17000365.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1700153777.hg.1 ENST00000384882.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700153777.hg.1 NR_029710.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700153777.hg.1 TC1700007485.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700153777.hg.1 NR_029710.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700153777.hg.1 ENST00000384882.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007485.hg.1 ENST00000384882.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007485.hg.1 NR_029710.1 Refseq
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3308971 NR_029710.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3308971 NR_029710.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308971 NR_029710.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308971 NR_029710.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0