Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274065_s1
          See other LOC100499194 GE Assays ›
          Gene Symbol:
          LOC100499194
          Gene Name:
          uncharacterized LOC100499194
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 113977669 - 113979984 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04274065_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100499194
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_034130.1 1 2222 85
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA HY226350.1 - 1 Chr. 2 - 113977670 - 113978085 [+] on Build GRCh38 85
          GenBankmRNA HY231638.1 - 1 Chr. 2 - 113977666 - 113977994 [+] on Build GRCh38 85
          GenBankmRNA HY223781.1 - 1 Chr. 2 - 113977666 - 113978124 [+] on Build GRCh38 85
          GenBankmRNA HY235400.1 - 1 Chr. 2 - 113977670 - 113978127 [+] on Build GRCh38 85
          GenBankmRNA BF980090.1 - 1 Chr. 2 - 113977140 - 113977974 [+] on Build GRCh38 85
          GenBankmRNA HY244770.1 - 1 Chr. 2 - 113977670 - 113978141 [+] on Build GRCh38 85
          GenBankmRNA HY217529.1 - 1 Chr. 2 - 113977660 - 113978179 [+] on Build GRCh38 85
          GenBankmRNA AA705799.1 - 1 Chr. 2 - 113977669 - 113978095 [+] on Build GRCh38 85
          GenBankmRNA HY212355.1 - 1 Chr. 2 - 113977670 - 113978103 [+] on Build GRCh38 85
          GenBankmRNA HY207987.1 - 1 Chr. 2 - 113977661 - 113978178 [+] on Build GRCh38 85
          GenBankmRNA HY231113.1 - 1 Chr. 2 - 113977674 - 113978122 [+] on Build GRCh38 85
          GenBankmRNA DB525744.1 - 1 Chr. 2 - 113977669 - 113978086 [-] on Build GRCh38 85
          GenBankmRNA DB519296.1 - 1 Chr. 2 - 113977666 - 113978095 [-] on Build GRCh38 85
          GenBankmRNA HY204260.1 - 1 Chr. 2 - 113977666 - 113978149 [+] on Build GRCh38 85
          GenBankmRNA HY190162.1 - 1 Chr. 2 - 113977670 - 113978159 [+] on Build GRCh38 85
          GenBankmRNA HY188262.1 - 1 Chr. 2 - 113977666 - 113978235 [+] on Build GRCh38 85
          GenBankmRNA AI205815.1 - 1 Chr. 2 - 113977669 - 113977989 [+] on Build GRCh38 85
          GenBankmRNA HY236558.1 - 1 Chr. 2 - 113977660 - 113978127 [+] on Build GRCh38 85
          GenBankmRNA HY189566.1 - 1 Chr. 2 - 113977660 - 113978148 [+] on Build GRCh38 85

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream long intergenic non-protein coding RNA 1191 LINC01191 VIN,
          lnc-ACTR3
          Anti-sense Chr. 2 - 113979569 - 114007310 on NCBI Build GRCh38
          Downstream dipeptidyl peptidase like 10 DPP10 DPL2,
          DPPY,
          DPRP-3,
          DPRP3
          462.3kb Anti-sense Chr. 2 - 114442358 - 115845752 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200013957.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02002233.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02036214.hg.1 NR_034130.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02036214.hg.1 TC02002233.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200202396.hg.1 NR_034130.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200202396.hg.1 TC0200013957.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200202396.hg.1 NR_034130.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200202397.hg.1 NR_034130.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200202397.hg.1 TC0200013957.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200202397.hg.1 NR_034130.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200202395.hg.1 NR_034130.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200202395.hg.1 TC0200013957.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200202395.hg.1 NR_034130.1 Refseq
          Human Genome U95 Set 51660_at - -
          Human Genome U95 Set 51660_PM_at - -
          Human Genome U95 Set 84790_g_at - -
          Human Genome U95 Set 84788_at - -
          Human Genome U95 Set 84788_PM_at - -
          Human Genome U95 Set 84790_PM_g_at - -
          Human Genome U95 Set 79529_PM_at - -
          Human Genome U95 Set 79529_at - -
          Human Genome U133 Set 231637_at - -
          Human Genome U133 Set 231637_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 153805 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-8696d955b-mh7vc:80/100.66.135.11:80.
          git-commit: b6496d4449ebd8b46e03b29da6eff4841d0bfc13
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline