Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16015545.hg.1 |
NR_038233.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16015545.hg.1 |
TC16001113.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16015546.hg.1 |
NR_038233.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16015546.hg.1 |
TC16001113.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010309.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010309.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010309.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010309.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010309.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010309.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010309.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010309.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010309.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010309.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010309.hg.1 |
ENST00000565742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010309.hg.1 |
ENST00000565742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010309.hg.1 |
ENST00000565742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010309.hg.1 |
ENST00000565742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010309.hg.1 |
NR_038233.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010309.hg.1 |
NR_038233.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010309.hg.1 |
NR_038233.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010309.hg.1 |
NR_038233.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010309.hg.1 |
NR_038233.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600179189.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600179189.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600179189.hg.1 |
ENST00000565742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600179189.hg.1 |
NR_038233.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600179189.hg.1 |
TC1600010309.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600179189.hg.1 |
NR_038233.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600179189.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600179189.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600179189.hg.1 |
ENST00000565742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010309.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010309.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010309.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010309.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010309.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010309.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010309.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010309.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010309.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010309.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010309.hg.1 |
ENST00000565742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010309.hg.1 |
ENST00000565742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010309.hg.1 |
ENST00000565742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010309.hg.1 |
ENST00000565742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010309.hg.1 |
NR_038233.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010309.hg.1 |
NR_038233.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010309.hg.1 |
NR_038233.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010309.hg.1 |
NR_038233.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010309.hg.1 |
NR_038233.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600179188.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600179188.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600179188.hg.1 |
NR_038233.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600179188.hg.1 |
TC1600010309.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600179188.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600179188.hg.1 |
ENST00000561513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600179188.hg.1 |
NR_038233.1 |
Refseq |