Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274314_m1
          See other LOC100507194 GE Assays ›
          Gene Symbol:
          LOC100507194
          Gene Name:
          uncharacterized LOC100507194
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 3182817 - 3195767 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04274314_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100507194
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038295.1 2 - 3 535 108
          NR_038296.1 2 - 3 508 108
          GenBank mRNA BC027024.1 - 2 - 3 525 108
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DT217643.1 - 2 - 3 457 Chr. 6 - 3183466 - 3195715 [-] on Build GRCh38 108
          GenBankmRNA BI089937.1 - 2 - 3 525 Chr. 6 - 3183350 - 3195757 [-] on Build GRCh38 108

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream tubulin beta 2A class IIa TUBB2A CDCBM5,
          TUBB,
          TUBB2
          -25.3kb Sense Chr. 6 - 3153666 - 3157549 on NCBI Build GRCh38
          Downstream tubulin beta 2B class IIb TUBB2B PMGYSA,
          bA506K6.1
          33.0kb Sense Chr. 6 - 3224261 - 3227734 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600010569.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06001227.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515542 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515572 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515521 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515549 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515544 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515569 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515507 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515551 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24702254 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515534 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515581 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24702229 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24702231 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24702255 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515520 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24702256 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15778718 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515554 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24702225 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515508 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515565 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24702230 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515570 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515528 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24702226 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515547 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15778717 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15778719 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515548 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515538 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515522 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515545 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515562 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515574 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515550 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515523 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24702261 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24702232 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515580 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515506 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515578 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24702184 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24702263 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515571 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515536 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24702241 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515567 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04515559 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24702266 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 5080 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 60858 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 369574 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 535622 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 957450 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 147543 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P17428532 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P17428532 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 49266 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 24061 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P24733965 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17428532 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17428538 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P24734346 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01411349 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P24732921 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P24734346 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P17428532 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR06014570.hg.1 ENST00000425384.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06014570.hg.1 ENST00000435043.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06014570.hg.1 NR_038295.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06014570.hg.1 NR_038296.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06014570.hg.1 TC06001227.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06014568.hg.1 ENST00000425384.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06014568.hg.1 ENST00000435043.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06014568.hg.1 NR_038295.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06014568.hg.1 NR_038296.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06014568.hg.1 TC06001227.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06014572.hg.1 ENST00000425384.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06014572.hg.1 ENST00000435043.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06014572.hg.1 NR_038295.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06014572.hg.1 NR_038296.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06014572.hg.1 TC06001227.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 ENST00000435043.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 ENST00000435043.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 ENST00000435043.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 ENST00000435043.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 ENST00000435043.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 ENST00000435043.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 NR_038295.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 NR_038295.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 NR_038295.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 NR_038295.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 NR_038295.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 NR_038296.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 NR_038296.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 NR_038296.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010569.hg.1 NR_038296.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06014571.hg.1 ENST00000435043.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06014571.hg.1 NR_038295.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06014571.hg.1 TC06001227.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0600171811.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171811.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171811.hg.1 ENST00000435043.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171811.hg.1 NR_038295.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600171811.hg.1 NR_038296.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600171811.hg.1 TC0600010569.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600171811.hg.1 NR_038295.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600171811.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171811.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171811.hg.1 ENST00000435043.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171811.hg.1 NR_038296.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600171817.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171817.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171817.hg.1 ENST00000435043.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171817.hg.1 NR_038295.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600171817.hg.1 NR_038296.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600171817.hg.1 TC0600010569.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600171817.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171817.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171817.hg.1 ENST00000435043.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171817.hg.1 NR_038295.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600171817.hg.1 NR_038296.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600171816.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171816.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171816.hg.1 ENST00000435043.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171816.hg.1 NR_038295.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600171816.hg.1 NR_038296.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600171816.hg.1 TC0600010569.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600171816.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171816.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171816.hg.1 ENST00000435043.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171816.hg.1 NR_038295.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600171816.hg.1 NR_038296.1 Refseq
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 ENST00000435043.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 ENST00000435043.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 ENST00000435043.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 ENST00000435043.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 ENST00000435043.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 ENST00000435043.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 NR_038295.1 Refseq
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 NR_038295.1 Refseq
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 NR_038295.1 Refseq
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 NR_038295.1 Refseq
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 NR_038295.1 Refseq
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 NR_038296.1 Refseq
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 NR_038296.1 Refseq
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 NR_038296.1 Refseq
          Affymetrix Human Clariom D Assay TC0600010569.hg.1 NR_038296.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600171820.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171820.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171820.hg.1 ENST00000435043.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171820.hg.1 NR_038295.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600171820.hg.1 NR_038296.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600171820.hg.1 TC0600010569.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600171820.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171820.hg.1 ENST00000425384.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171820.hg.1 ENST00000435043.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171820.hg.1 NR_038295.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600171820.hg.1 NR_038296.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600171819.hg.1 ENST00000435043.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171819.hg.1 NR_038295.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600171819.hg.1 TC0600010569.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600171819.hg.1 ENST00000435043.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171819.hg.1 NR_038295.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A055215 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A055215 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix