Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274330_m1
          See other LINC01364 GE Assays ›
          Gene Symbol:
          LINC01364
          Gene Name:
          long intergenic non-protein coding RNA 1364
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 87353524 - 87371655 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04274330_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC01364
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038324.1 2 - 3 193 90
          GenBank mRNA BC043534.1 - 2 - 3 193 90
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BI832138.1 - 2 - 3 202 Chr. 1 - 87353685 - 87371655 [-] on Build GRCh38 90
          GenBankmRNA BX103394.1 - 2 - 3 169 Chr. 1 - 87354931 - 87371631 [-] on Build GRCh38 90

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream LIM domain only 4 LMO4 -5.9kb Anti-sense Chr. 1 - 87328468 - 87348923 on NCBI Build GRCh38
          Downstream PKN2 antisense RNA 1 PKN2-AS1 1.2mb Sense Chr. 1 - 88537513 - 88685204 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100014823.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002838.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385055 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385137 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15082036 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385132 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385082 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355937 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355963 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385097 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385109 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385101 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385049 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385140 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385087 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385075 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385066 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385084 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355953 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385088 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385061 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355970 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355945 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385057 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385053 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355952 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355930 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385115 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385116 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385112 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385059 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15082040 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385134 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355925 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385083 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385121 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355949 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355922 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355926 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355910 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355938 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385056 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355987 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385070 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355920 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385131 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15082039 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355927 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15082041 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355942 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385108 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355993 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355912 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385086 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15082038 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385095 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385077 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355980 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355962 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15082033 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385054 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355928 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385117 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355985 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385072 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385123 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385114 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385050 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385103 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20385139 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355979 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00355909 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 34580 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 81567 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 609321 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 348641 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 789731 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 401449 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 39393 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 48129 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P00115401 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P00115383 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 12476 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00115395 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15203776 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00115383 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00115388 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15203751 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00115401 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P10214706 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P10217622 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P15203776 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00115383 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00115401 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01044251.hg.1 ENST00000452509.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01044251.hg.1 NR_038324.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01044251.hg.1 TC01002838.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01044252.hg.1 ENST00000452509.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01044252.hg.1 NR_038324.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01044252.hg.1 TC01002838.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100014823.hg.1 ENST00000452509.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014823.hg.1 ENST00000452509.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014823.hg.1 ENST00000452509.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014823.hg.1 ENST00000452509.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014823.hg.1 NR_038324.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014823.hg.1 NR_038324.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014823.hg.1 NR_038324.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014823.hg.1 NR_038324.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100210406.hg.1 ENST00000452509.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100210406.hg.1 NR_038324.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100210406.hg.1 TC0100014823.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100210406.hg.1 NR_038324.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100210406.hg.1 ENST00000452509.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100210407.hg.1 ENST00000452509.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100210407.hg.1 NR_038324.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100210407.hg.1 TC0100014823.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100210407.hg.1 ENST00000452509.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100210407.hg.1 NR_038324.1 Refseq
          Affymetrix Human Clariom D Assay TC0100014823.hg.1 ENST00000452509.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014823.hg.1 ENST00000452509.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014823.hg.1 ENST00000452509.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014823.hg.1 ENST00000452509.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014823.hg.1 NR_038324.1 Refseq
          Affymetrix Human Clariom D Assay TC0100014823.hg.1 NR_038324.1 Refseq
          Affymetrix Human Clariom D Assay TC0100014823.hg.1 NR_038324.1 Refseq
          Affymetrix Human Clariom D Assay TC0100014823.hg.1 NR_038324.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047877 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047877 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1906957 BC043534.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1906957 NR_038324.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1906957 BC043534.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1906957 NR_038324.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1906957 BC043534.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1906957 NR_038324.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip ILMN_76100 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_76100 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1906957 BC043534.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1906957 NR_038324.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1906957 BC043534.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1906957 NR_038324.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1906957 BC043534.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1906957 NR_038324.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0